Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ADM SIRNA Assays ›
Gene Symbol:
ADM
Gene Name:
adrenomedullin
Gene Aliases:
AM, PAMP
Chromosome Location:
Chr.11: 10304980 - 10307402 on Build GRCh38
UniGene ID:
Hs.441047
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
9524
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ADM
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001124.2
NP_001115.1
4
1090
GenBank mRNA
BC015961.2
-
4
985
3506276
D14874.1
-
4
975
Back To Top
More Information
Panther Classification:
Molecular Function -
peptide hormone
intercellular signal molecule
Gene Ontology Categories:
Function(s)
Process(es)
vasculogenesis
response to hypoxia
neural tube closure
response to yeast
regulation of the force of heart contraction
G-protein coupled receptor internalization
cAMP biosynthetic process
progesterone biosynthetic process
signal transduction
positive regulation of cytosolic calcium ion concentration
cell-cell signaling
heart development
female pregnancy
aging
blood circulation
androgen metabolic process
positive regulation of cell proliferation
negative regulation of cell proliferation
response to cold
response to wounding
positive regulation of heart rate
antibacterial humoral response
antifungal humoral response
cAMP-mediated signaling
positive regulation of cAMP biosynthetic process
organ regeneration
neuron projection regeneration
receptor internalization
response to lipopolysaccharide
response to insulin
odontogenesis of dentin-containing tooth
response to starvation
positive regulation of apoptotic process
negative regulation of vascular permeability
positive regulation of angiogenesis
negative regulation of vasoconstriction
positive regulation of vasodilation
hormone secretion
developmental growth
defense response to Gram-negative bacterium
defense response to Gram-positive bacterium
response to glucocorticoid
branching involved in labyrinthine layer morphogenesis
spongiotrophoblast layer development
vascular smooth muscle cell development
positive regulation of vasculogenesis
receptor binding
hormone activity
protein binding
adrenomedullin receptor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0