Search
Search

Atg4c| Gene Symbol: Atg4c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145967.1 | NP_001139439.1 | 6 | 967 | ||
| NM_175029.3 | NP_778194.3 | 6 | 1349 | |||
| GenBank mRNA | AK134838.1 | - | 6 | 1116 | ||
| AK156463.1 | - | 6 | 1119 | |||
| AK170747.1 | - | 6 | 1106 | |||
| BC058981.1 | - | 6 | 1349 | 6827140 | ||