Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other AGER SIRNA Assays ›
          Gene Symbol:
          AGER
          Gene Name:
          advanced glycosylation end product-specific receptor
          Gene Aliases:
          RAGE, SCARJ1
          Chromosome Location:
          Chr.6: 32180968 - 32185247 on Build GRCh38
          UniGene ID:
          Hs.534342
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 9627

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: AGER
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001136.4 NP_001127.1 5, 6 615
          NM_001206929.1 NP_001193858.1 5, 6 663
          NM_001206932.1 NP_001193861.1 5, 6 573
          NM_001206934.1 NP_001193863.1 5, 6 663
          NM_001206936.1 NP_001193865.1 5, 6 615
          NM_001206940.1 NP_001193869.1 5, 6 615
          NM_001206954.1 NP_001193883.1 5, 6 615
          NM_001206966.1 NP_001193895.1 5, 6 615
          NM_172197.2 NP_751947.1 5, 6 573
          NR_038190.1 4, 5 798
          XM_017010328.1 XP_016865817.1 5, 6 1588
          GenBank mRNA AB036432.1 - 5, 6 539
          AB061668.1 - 5, 6 539
          AB061669.1 - 4, 5 722
          AJ133822.1 - 5, 6 488
          AK298100.1 - 5, 6 617
          AK313178.1 - 5, 6 543
          AY755619.1 - 5, 6 525
          AY755620.1 - 5, 6 525
          AY755621.1 - 5, 6 573
          AY755622.1 - 5, 6 573
          AY755623.1 - 5, 6 525
          AY755624.1 - 5, 6 483
          AY755628.1 - 5, 6 525
          BC020669.1 - 5, 6 534 4718076
          DQ104251.1 - 5, 6 525
          DQ104252.1 - 5, 6 483
          DQ104253.1 - 5, 6 525
          DQ104254.1 - 4, 5 708
          EU117141.1 - 5, 6 525
          EU428812.1 - 4, 5 698
          EU428813.1 - 4, 5 698
          EU428815.1 - 4, 5 698
          EU428816.1 - 4, 5 698
          EU826618.1 - 5, 6 515
          EU826619.1 - 5, 6 563
          FJ545655.1 - 5, 6 587
          GU139370.1 - 2, 3 973
          GU139371.1 - 2, 3 972
          GU139372.1 - 4, 5 772
          GU139373.1 - 3, 4 903
          GU139374.1 - 3, 4 849
          GU139375.1 - 4, 5 772
          GU139377.1 - 4, 5 721
          GU139378.1 - 5, 6 537
          HQ148133.1 - 5, 6 539
          HQ148135.1 - 4, 5 719
          KC692917.1 - 5, 6 515
          M91211.1 - 5, 6 515

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          cell adhesion molecule immunoglobulin superfamily cell adhesion molecule

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of T cell mediated cytotoxicity
          negative regulation of protein phosphorylation
          positive regulation of protein phosphorylation
          negative regulation of endothelial cell proliferation
          inflammatory response
          negative regulation of cell adhesion
          cell surface receptor signaling pathway
          JAK-STAT cascade
          brain development
          glycoprotein metabolic process
          response to wounding
          response to fructose
          positive regulation of autophagy
          negative regulation of endothelial cell migration
          positive regulation of gene expression
          positive regulation of epithelial to mesenchymal transition
          positive regulation of fibroblast migration
          response to activity
          positive regulation of smooth muscle cell migration
          lung development
          neuron projection development
          negative regulation of interleukin-10 production
          positive regulation of interleukin-12 production
          negative regulation of collagen biosynthetic process
          response to vitamin A
          response to genistein
          negative regulation of osteoblast proliferation
          cellular response to drug
          positive regulation of activated T cell proliferation
          positive regulation of neuron apoptotic process
          innate immune response
          positive regulation of fibroblast proliferation
          positive regulation of smooth muscle cell proliferation
          positive regulation of inflammatory response
          positive regulation of NF-kappaB transcription factor activity
          response to methylglyoxal
          calcium ion homeostasis
          response to hyperoxia
          positive regulation of phagocytosis, engulfment
          transdifferentiation
          cellular response to hydrogen peroxide
          cellular response to glucose stimulus
          cellular response to fatty acid
          cellular response to organic cyclic compound
          response to selenite ion
          positive regulation of potassium ion transmembrane transporter activity
          positive regulation of endothelial cell apoptotic process
          positive regulation of reactive oxygen species metabolic process
          regulation of CD4-positive, alpha-beta T cell activation
          positive regulation of type B pancreatic cell apoptotic process
          positive regulation of dendritic cell differentiation
          receptor activity
          transmembrane signaling receptor activity
          protein binding
          identical protein binding
          S100 protein binding
          advanced glycation end-product receptor activity
          high mobility group box 1 binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-4x8c5:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0