Search
Search

ANPEP| Gene Symbol: ANPEP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001150.2 | NP_001141.2 | 12 | 2094 | ||
| XM_005254892.4 | XP_005254949.1 | 12 | 2102 | |||
| XM_011521473.1 | XP_011519775.1 | 12 | 2245 | |||
| GenBank mRNA | AB209918.1 | - | 12 | 2090 | ||
| AK298134.1 | - | 6 | 564 | |||
| AK298242.1 | - | 6 | 615 | |||
| AK300946.1 | - | 8 | 825 | |||
| BC058928.1 | - | 11 | 1895 | 6020902 | ||
| M22324.1 | - | 11 | 1922 | |||