Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DLG2 SIRNA Assays ›
Gene Symbol:
DLG2
Gene Name:
discs large MAGUK scaffold protein 2
Gene Aliases:
PPP1R58, PSD-93, PSD93, chapsyn-110
Chromosome Location:
Chr.11: 83455009 - 85628534 on Build GRCh38
UniGene ID:
Hs.367656
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
9920
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DLG2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001142699.1
NP_001136171.1
28
3322
NM_001142700.1
NP_001136172.1
20
2357
NM_001142702.1
NP_001136174.1
11
1399
NM_001206769.1
NP_001193698.1
22
3006
NM_001300983.1
NP_001287912.1
23
2944
NM_001364.3
NP_001355.2
23
2998
XM_005273810.4
XP_005273867.1
22
2641
XM_005273811.4
XP_005273868.1
22
2587
XM_011544778.2
XP_011543080.1
29
4613
XM_011544780.2
XP_011543082.1
29
4559
XM_011544782.2
XP_011543084.1
24
3084
XM_011544788.2
XP_011543090.1
21
2431
XM_017017254.1
XP_016872743.1
30
4655
XM_017017255.1
XP_016872744.1
29
4616
XM_017017256.1
XP_016872745.1
29
4145
XM_017017257.1
XP_016872746.1
28
5077
XM_017017258.1
XP_016872747.1
27
4253
XM_017017259.1
XP_016872748.1
27
3974
XM_017017260.1
XP_016872749.1
26
4906
XM_017017261.1
XP_016872750.1
28
4403
XM_017017262.1
XP_016872751.1
23
3108
XM_017017263.1
XP_016872752.1
25
3126
XM_017017264.1
XP_016872753.1
24
3303
XM_017017265.1
XP_016872754.1
24
3300
XM_017017266.1
XP_016872755.1
22
3010
XM_017017267.1
XP_016872756.1
24
3208
XM_017017268.1
XP_016872757.1
24
3029
XM_017017269.1
XP_016872758.1
24
3730
XM_017017270.1
XP_016872759.1
24
3030
XM_017017271.1
XP_016872760.1
23
3207
XM_017017272.1
XP_016872761.1
23
3204
XM_017017273.1
XP_016872762.1
22
2959
XM_017017274.1
XP_016872763.1
24
3154
XM_017017275.1
XP_016872764.1
21
2909
XM_017017276.1
XP_016872765.1
23
3632
XM_017017277.1
XP_016872766.1
23
2952
XM_017017278.1
XP_016872767.1
21
2856
XM_017017279.1
XP_016872768.1
23
2874
XM_017017280.1
XP_016872769.1
23
3551
XM_017017281.1
XP_016872770.1
22
3051
XM_017017283.1
XP_016872772.1
22
2856
XM_017017284.1
XP_016872773.1
22
2528
XM_017017285.1
XP_016872774.1
22
3478
XM_017017286.1
XP_016872775.1
20
2392
XM_017017287.1
XP_016872776.1
13
1412
XM_017017288.1
XP_016872777.1
12
1251
XM_017017289.1
XP_016872778.1
12
2826
XM_017017290.1
XP_016872779.1
12
1471
XM_017017291.1
XP_016872780.1
11
1218
XM_017017292.1
XP_016872781.1
11
2730
GenBank mRNA
AK126776.1
-
20
2357
AK294375.1
-
22
2475
AK299778.1
-
12
1410
AK299797.1
-
24
2744
CR749820.1
-
28
3322
CR933674.1
-
11
1368
U32376.1
-
24
2971
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_022282, XM_008759648, XM_008759649, U49049.
This siRNA also targets the following transcripts in Mouse: NM_011807, NM_001243047, NM_001243046, AK039754, AK030221.
Panther Classification:
Molecular Function -
scaffold/adaptor protein
Gene Ontology Categories:
Function(s)
Process(es)
chemical synaptic transmission
nervous system development
negative regulation of phosphatase activity
sensory perception of pain
receptor clustering
establishment or maintenance of epithelial cell apical/basal polarity
GMP metabolic process
GDP metabolic process
receptor localization to synapse
guanylate kinase activity
protein binding
kinase binding
ionotropic glutamate receptor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-4sktr:80/100.66.76.202:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0