Search
Search

Hist1h2be| Gene Symbol: Hist1h2be | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177653.1 | NP_001171124.1 | 2 | 793 | ||
| NM_178194.4 | NP_835501.1 | 2 | 819 | |||
| GenBank mRNA | AK030546.1 | - | 2 | 821 | ||
| AK049948.1 | - | 2 | 794 | |||
| BC069889.1 | - | 1 | 607 | 4217814 | ||