Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ACTN4 SIRNA Assays ›
          Gene Symbol:
          ACTN4
          Gene Name:
          actinin alpha 4
          Gene Aliases:
          ACTININ-4, FSGS, FSGS1
          Chromosome Location:
          Chr.19: 38647616 - 38730532 on Build GRCh38
          UniGene ID:
          Hs.270291
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS100124
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ACTN4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001322033.1 NP_001308962.1 3 469
          NM_004924.5 NP_004915.2 3 469
          XM_005259281.4 XP_005259338.1 3 469
          XM_006723406.2 XP_006723469.1 3 469
          XM_017027331.1 XP_016882820.1 3 469
          GenBank mRNA AK299955.1 - 3 398
          AK304554.1 - 3 398
          AK309361.1 - 3 398
          AK309363.1 - 3 436
          AK309364.1 - 3 398
          AU118403.1 - 3 398
          BC005033.1 - 3 352 3842046
          BP332596.1 - 3 458
          D89980.1 - 3 346
          HY048627.1 - 3 470
          KU177881.1 - 3 339
          U48734.1 - 3 413

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          response to hypoxia
          platelet degranulation
          protein transport
          vesicle transport along actin filament
          positive regulation of cell migration
          positive regulation of sodium:proton antiporter activity
          peroxisome proliferator activated receptor signaling pathway
          regulation of apoptotic process
          retinoic acid receptor signaling pathway
          positive regulation of pinocytosis
          actin filament bundle assembly
          negative regulation of cellular component movement
          positive regulation of cellular component movement
          bicellular tight junction assembly
          negative regulation of substrate adhesion-dependent cell spreading
          positive regulation of NIK/NF-kappaB signaling
          protein localization to bicellular tight junction
          regulation of nucleic acid-templated transcription
          RNA polymerase II regulatory region sequence-specific DNA binding
          nucleoside binding
          actin binding
          integrin binding
          calcium ion binding
          protein binding
          ligand-dependent nuclear receptor transcription coactivator activity
          chromatin DNA binding
          nuclear hormone receptor binding
          protein homodimerization activity
          retinoic acid receptor binding
          ion channel binding
          poly(A) RNA binding
          protein N-terminus binding
          actin filament binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0