Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ACVRL1 SIRNA Assays ›
          Gene Symbol:
          ACVRL1
          Gene Name:
          activin A receptor like type 1
          Gene Aliases:
          ACVRLK1, ALK-1, ALK1, HHT, HHT2, ORW2, SKR3, TSR-I
          Chromosome Location:
          Chr.12: 51906913 - 51923361 on Build GRCh38
          UniGene ID:
          Hs.591026
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS100152
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ACVRL1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000020.2 NP_000011.2 10 1779
          NM_001077401.1 NP_001070869.1 9 1642
          XM_005269235.2 XP_005269292.1 11 1781
          XM_011539008.1 XP_011537310.1 7 1325
          GenBank mRNA AK298493.1 - 7 806
          AK300619.1 - 5 760
          AK303331.1 - 6 1037
          BC042637.1 - 9 1528 5179243
          L17075.1 - 9 1587
          Z22533.1 - 10 1778

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          serine/threonine protein kinase receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          angiogenesis
          in utero embryonic development
          regulation of endothelial cell proliferation
          negative regulation of endothelial cell proliferation
          positive regulation of endothelial cell proliferation
          lymphangiogenesis
          blood vessel maturation
          blood vessel remodeling
          blood vessel endothelial cell proliferation involved in sprouting angiogenesis
          endocardial cushion morphogenesis
          regulation of DNA replication
          regulation of transcription, DNA-templated
          protein phosphorylation
          negative regulation of cell adhesion
          signal transduction
          transforming growth factor beta receptor signaling pathway
          blood circulation
          regulation of blood pressure
          negative regulation of cell proliferation
          negative regulation of endothelial cell migration
          negative regulation of gene expression
          positive regulation of pathway-restricted SMAD protein phosphorylation
          signal transduction by protein phosphorylation
          negative regulation of cell growth
          negative regulation of cell migration
          BMP signaling pathway
          positive regulation of BMP signaling pathway
          positive regulation of chondrocyte differentiation
          activin receptor signaling pathway
          wound healing, spreading of epidermal cells
          dorsal aorta morphogenesis
          regulation of blood vessel endothelial cell migration
          negative regulation of blood vessel endothelial cell migration
          negative regulation of endothelial cell differentiation
          positive regulation of endothelial cell differentiation
          positive regulation of angiogenesis
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          protein heterooligomerization
          negative regulation of focal adhesion assembly
          lymphatic endothelial cell differentiation
          artery development
          venous blood vessel development
          endothelial tube morphogenesis
          retina vasculature development in camera-type eye
          cellular response to transforming growth factor beta stimulus
          cellular response to BMP stimulus
          endocardial cushion to mesenchymal transition
          negative regulation of DNA biosynthetic process
          protein serine/threonine kinase activity
          transmembrane receptor protein serine/threonine kinase activity
          receptor signaling protein serine/threonine kinase activity
          transforming growth factor beta-activated receptor activity
          transforming growth factor beta receptor activity, type I
          protein binding
          ATP binding
          activin receptor activity, type I
          protein kinase binding
          SMAD binding
          metal ion binding
          activin binding
          transforming growth factor beta binding
          BMP receptor activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0