Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other BDNF SIRNA Assays ›
Gene Symbol:
BDNF
Gene Name:
brain derived neurotrophic factor
Gene Aliases:
ANON2, BULN2
Chromosome Location:
Chr.11: 27654893 - 27722058 on Build GRCh38
UniGene ID:
Hs.502182
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS100994
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: BDNF
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001143805.1
NP_001137277.1
2
841
NM_001143806.1
NP_001137278.1
2
1056
NM_001143807.1
NP_001137279.1
2
942
NM_001143808.1
NP_001137280.1
2
778
NM_001143809.1
NP_001137281.1
2
788
NM_001143810.1
NP_001137282.1
3
1071
NM_001143811.1
NP_001137283.1
4
1188
NM_001143812.1
NP_001137284.1
2
930
NM_001143813.1
NP_001137285.1
2
1012
NM_001143814.1
NP_001137286.1
3
1137
NM_001143816.1
NP_001137288.1
2
1535
NM_001709.4
NP_001700.2
2
1030
NM_170731.4
NP_733927.1
2
1352
NM_170732.4
NP_733928.1
2
1139
NM_170733.3
NP_733929.1
2
1042
NM_170734.3
NP_733930.1
2
990
NM_170735.5
NP_733931.1
1
1769
XM_011520280.2
XP_011518582.1
2
1936
GenBank mRNA
AF400438.1
-
1
702
AK289763.1
-
2
838
AK289853.1
-
2
739
AY054392.2
-
2
1043
AY054393.2
-
2
785
AY054394.2
-
2
1000
AY054395.2
-
2
1083
AY054396.2
-
2
841
AY054397.2
-
2
924
AY054398.2
-
2
971
AY054399.2
-
3
856
AY054400.2
-
2
957
AY054406.3
-
3
681
AY656701.1
-
1
684
BC029795.1
-
2
1015
5193877
EF674517.1
-
2
841
EF674518.1
-
2
1056
EF674519.1
-
2
1139
EF674520.1
-
2
942
EF674521.1
-
2
1042
EF689009.1
-
1
1783
EF689010.1
-
2
1549
EF689011.1
-
2
778
EF689012.1
-
2
788
EF689013.1
-
2
930
EF689014.1
-
2
1012
EF689015.1
-
2
1030
EF689016.1
-
3
1137
EF689017.1
-
3
889
EF689018.1
-
2
990
EF689019.1
-
3
1071
EF689020.1
-
4
1188
EF689021.1
-
2
1352
KP229412.1
-
1
684
KU177943.1
-
1
684
KU177944.1
-
2
567
KU177945.1
-
2
540
M61176.1
-
1
1246
X60201.1
-
2
980
X91251.1
-
2
968
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: AY176065.
This siRNA also targets the following transcripts in Mouse: NM_001285420, NM_001285419, NM_001285417, NM_001285421, NM_001048139, and others.
Panther Classification:
Molecular Function -
neurotrophic factor
Gene Ontology Categories:
Function(s)
Process(es)
cell-cell signaling
nervous system development
axon guidance
synapse assembly
brain-derived neurotrophic factor receptor signaling pathway
positive regulation of brain-derived neurotrophic factor receptor signaling pathway
negative regulation of neuron apoptotic process
collateral sprouting
positive regulation of collateral sprouting
positive regulation of synapse assembly
neurotrophin TRKB receptor binding
growth factor activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-nwsk8:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0