Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CAV2 SIRNA Assays ›
          Gene Symbol:
          CAV2 CAV1 LOC105375463
          Gene Name:
          caveolin 2
          caveolin 1
          uncharacterized LOC105375463
          Gene Aliases:
          CAV
          BSCL3, CGL3, LCCNS, MSTP085, PPH3, VIP21
          -
          Chromosome Location:
          Chr.7: 116499601 - 116508541 on Build GRCh38
          Chromosome Location:
          Chr.7: 116524785 - 116561185 on Build GRCh38
          Chromosome Location:
          Chr.7: 116284417 - 116351782 on Build GRCh38
          UniGene ID:
          Hs.212332
          Hs.74034
          Hs.212332
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          siRNA ID HSS101398
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CAV2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001206747.1 NP_001193676.1 3 563
          NM_001206748.1 NP_001193677.1 2 448
          NM_001233.4 NP_001224.1 3 563
          NM_198212.2 NP_937855.1 2 375
          GenBank mRNA AA883287.1 - 2 193
          AB451285.1 - 3 382
          AB451417.1 - 3 382
          AF035752.1 - 3 402
          AK310786.1 - 2 236
          AK311964.1 - 3 405
          AY353255.1 - 3 394
          BC005256.1 - 3 563 3829012
          BT007051.1 - 3 382
          CR407685.1 - 3 382
          DA882274.1 - 2 448
          DQ656068.1 - 14 1844
          Gene Symbol: CAV1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA EF179102.1 - 7 709
          Gene Symbol: LOC105375463
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA EF070114.1 - 11 1084

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of endothelial cell proliferation
          positive regulation of endothelial cell proliferation
          vesicle fusion
          mitochondrion organization
          endoplasmic reticulum organization
          regulation of mitotic nuclear division
          insulin receptor signaling pathway
          vesicle organization
          receptor-mediated endocytosis of virus by host cell
          negative regulation of transforming growth factor beta receptor signaling pathway
          positive regulation of MAPK cascade
          positive regulation of GTPase activity
          positive regulation by host of viral release from host cell
          positive regulation by host of viral process
          vesicle docking
          skeletal muscle fiber development
          protein oligomerization
          positive regulation of dopamine receptor signaling pathway
          caveola assembly
          protein binding
          protein kinase binding
          protein binding, bridging
          protein complex scaffold
          protein homodimerization activity
          protein heterodimerization activity
          scaffold protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0