Search
Search

CHRNA2| Gene Symbol: CHRNA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000742.3 | NP_000733.2 | 3 | 795 | ||
| NM_001282455.1 | NP_001269384.1 | 3 | 795 | |||
| XM_005273397.2 | XP_005273454.1 | 3 | 719 | |||
| XM_006716282.1 | XP_006716345.1 | 4 | 706 | |||
| XM_011544388.1 | XP_011542690.1 | 4 | 797 | |||
| XM_011544389.1 | XP_011542691.1 | 3 | 795 | |||
| XM_017013003.1 | XP_016868492.1 | 3 | 772 | |||
| XM_017013004.1 | XP_016868493.1 | 4 | 706 | |||
| GenBank mRNA | AK296348.1 | - | 3 | 350 | ||
| AK314787.1 | - | 3 | 713 | |||
| BC153866.1 | - | 2 | 221 | 40121686 | ||
| CD013901.1 | - | 2 | 361 | |||
| DA096824.1 | - | 2 | 187 | |||
| U62431.1 | - | 3 | 740 | |||
| Y16281.1 | - | 2 | 186 | |||