Search
Search

CHRNB4| Gene Symbol: CHRNB4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000750.3 | NP_000741.1 | 5 | 1258 | ||
| XM_011521186.2 | XP_011519488.1 | 7 | 1288 | |||
| XM_011521187.2 | XP_011519489.1 | 6 | 1195 | |||
| XM_011521190.2 | XP_011519492.1 | 4 | 1426 | |||
| XM_011521191.2 | XP_011519493.1 | 4 | 1006 | |||
| XM_011521192.2 | XP_011519494.1 | 5 | 1399 | |||
| XM_017021885.1 | XP_016877374.1 | 5 | 1353 | |||
| XM_017021886.1 | XP_016877375.1 | 5 | 3160 | |||
| XM_017021887.1 | XP_016877376.1 | 5 | 1162 | |||
| XM_017021888.1 | XP_016877377.1 | 5 | 1162 | |||
| XM_017021889.1 | XP_016877378.1 | 5 | 1162 | |||
| GenBank mRNA | BC096080.1 | - | 5 | 1261 | 40000733 | |
| BC096082.1 | - | 5 | 1261 | 40000737 | ||
| U48861.1 | - | 5 | 1266 | |||
| U62439.1 | - | 5 | 1232 | |||
| X68275.1 | - | 4 | 1006 | |||
| Y08416.1 | - | 5 | 1237 | |||