Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CLN3 SIRNA Assays ›
Gene Symbol:
CLN3
Gene Name:
CLN3, battenin
Gene Aliases:
BTS, JNCL
Chromosome Location:
Chr.16: 28466653 - 28492302 on Build GRCh38
UniGene ID:
Hs.534667
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS102001
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CLN3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000086.2
NP_000077.1
3
465
NM_001042432.1
NP_001035897.1
4
501
NM_001286104.1
NP_001273033.1
4
501
NM_001286105.1
NP_001273034.1
2
386
NM_001286109.1
NP_001273038.1
2
386
NM_001286110.1
NP_001273039.1
2
386
GenBank mRNA
AF015593.1
-
3
196
AF015594.1
-
2
157
AF015595.1
-
2
160
AF015596.1
-
2
115
AF015597.1
-
3
215
AF015598.1
-
3
247
AF015599.1
-
3
210
AF015600.1
-
2
169
AF015601.1
-
3
241
AF077956.1
-
3
142
AF077957.1
-
3
142
AF077958.1
-
3
142
AF077959.1
-
3
142
AF077960.1
-
3
142
AF077961.1
-
3
142
AF077962.1
-
3
142
AF077963.1
-
3
142
AF077964.1
-
3
142
AF077965.1
-
3
142
AF077966.1
-
3
142
AF077967.1
-
3
142
AF077968.1
-
3
142
AF077971.1
-
3
142
AF077972.1
-
3
142
AF077973.1
-
2
63
AF078168.1
-
2
348
AF078169.1
-
3
142
AK090709.1
-
5
1496
AK294070.1
-
2
310
AK294250.1
-
4
271
AK295500.1
-
3
389
AK297690.1
-
2
328
AK302027.1
-
4
241
AK302138.1
-
3
190
AK313002.1
-
4
273
BC002394.2
-
4
259
2820893
BC004433.1
-
4
259
2820893
BC111068.1
-
2
316
5764535
BP252176.1
-
4
281
DA552461.1
-
4
501
DA703317.1
-
4
501
DA869025.1
-
3
432
DB101039.1
-
4
501
U32680.1
-
3
279
Back To Top
More Information
Panther Classification:
Molecular Function -
membrane traffic protein
membrane trafficking regulatory protein
Gene Ontology Categories:
Function(s)
Process(es)
action potential
globoside metabolic process
cellular amino acid metabolic process
ceramide metabolic process
glucosylceramide metabolic process
galactosylceramide metabolic process
sphingomyelin metabolic process
receptor-mediated endocytosis
vacuolar transport
lysosome organization
lysosomal lumen acidification
associative learning
arginine transport
macroautophagy
negative regulation of macroautophagy
protein processing
protein catabolic process
ionotropic glutamate receptor signaling pathway
lysosomal lumen pH elevation
neurotransmitter metabolic process
amyloid precursor protein catabolic process
negative regulation of apoptotic process
negative regulation of catalytic activity
negative regulation of neuron apoptotic process
negative regulation of proteolysis
vesicle transport along microtubule
neuromuscular process controlling balance
regulation of cytosolic calcium ion concentration
membrane organization
autophagosome maturation
protein binding
unfolded protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0