Search
Search

CCR4| Gene Symbol: CCR4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005508.4 | NP_005499.1 | 2 | 791 | ||
| XM_017005687.1 | XP_016861176.1 | 2 | 838 | |||
| GenBank mRNA | AK292674.1 | - | 2 | 721 | ||
| BC069139.1 | - | 1 | 676 | 7216998 | ||
| BC071751.1 | - | 2 | 791 | 30415446 | ||
| BC074935.2 | - | 1 | 651 | 30915268 | ||