Search
Search

CPA2| Gene Symbol: CPA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001869.2 | NP_001860.2 | 7 | 696 | ||
| GenBank mRNA | AK293378.1 | - | 7 | 692 | ||
| BC007009.1 | - | 7 | 696 | 3949984 | ||
| BC014571.1 | - | 7 | 698 | 3950117 | ||
| BC015140.1 | - | 7 | 698 | 4040975 | ||
| BT007403.1 | - | 7 | 671 | |||
| BX106544.1 | - | 1 | 67 | |||
| KU177990.1 | - | 7 | 671 | |||
| KU177991.1 | - | 8 | 777 | |||
| KU177992.1 | - | 6 | 569 | |||
| KU177993.1 | - | 4 | 388 | |||
| U19977.1 | - | 7 | 675 | |||