Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CTNND1 SIRNA Assays ›
Gene Symbol:
CTNND1
TMX2-CTNND1
Gene Name:
catenin delta 1
TMX2-CTNND1 readthrough (NMD candidate)
Gene Aliases:
CAS, CTNND, P120CAS, P120CTN, p120, p120(CAS), p120(CTN)
-
Chromosome Location:
Chr.11: 57761762 - 57819180 on Build GRCh38
Chromosome Location:
Chr.11: 57712523 - 57819180 on Build GRCh38
UniGene ID:
Hs.166011
Hs.166011
Species:
Human
Species Specific ID (Flybase ID):
-
-
siRNA ID
HSS102463
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CTNND1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001085458.1
NP_001078927.1
6
1423
NM_001085459.1
NP_001078928.1
6
1423
NM_001085460.1
NP_001078929.1
6
1350
NM_001085461.1
NP_001078930.1
6
1423
NM_001085462.1
NP_001078931.1
5
1231
NM_001085463.1
NP_001078932.1
4
942
NM_001085464.1
NP_001078933.1
4
942
NM_001085465.1
NP_001078934.1
4
942
NM_001085466.1
NP_001078935.1
4
942
NM_001085467.1
NP_001078936.1
4
1015
NM_001085468.1
NP_001078937.1
4
942
NM_001085469.1
NP_001078938.1
3
870
NM_001206883.1
NP_001193812.1
5
1122
NM_001206884.1
NP_001193813.1
5
1122
NM_001206885.1
NP_001193814.1
6
1423
NM_001206886.1
NP_001193815.1
5
1122
NM_001206887.1
NP_001193816.1
5
1122
NM_001206888.1
NP_001193817.1
5
1122
NM_001206889.1
NP_001193818.1
6
1241
NM_001206890.1
NP_001193819.1
4
964
NM_001206891.1
NP_001193820.1
6
1314
NM_001331.2
NP_001322.1
6
1423
GenBank mRNA
AF062317.1
-
6
1388
AF062319.1
-
5
1087
AF062320.1
-
5
1087
AF062321.1
-
6
1388
AF062322.1
-
5
1087
AF062323.1
-
4
907
AF062324.1
-
6
1315
AF062325.1
-
5
1087
AF062327.1
-
5
1087
AF062328.1
-
6
1388
AF062329.1
-
6
1206
AF062330.1
-
4
929
AF062331.1
-
6
1279
AF062332.1
-
4
907
AF062334.1
-
4
980
AF062335.1
-
4
907
AF062336.1
-
3
835
AF062337.1
-
4
907
AF062338.1
-
4
907
AF062339.1
-
6
1388
AF062341.1
-
6
1388
AF062343.1
-
5
1196
AK292554.1
-
6
1365
BC010501.1
-
2
544
BC075795.1
-
6
1239
5269008
CR749275.1
-
6
1183
Gene Symbol: TMX2-CTNND1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NR_037646.1
7
1411
GenBank mRNA
AB002382.1
-
7
1317
Back To Top
More Information
Panther Classification:
Molecular Function -
intermediate filament binding protein
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
cell adhesion
brain development
Wnt signaling pathway
single organismal cell-cell adhesion
adherens junction organization
negative regulation of canonical Wnt signaling pathway
receptor binding
protein binding
protein kinase binding
cadherin binding
cadherin binding involved in cell-cell adhesion
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0