Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CTNND1 SIRNA Assays ›
Gene Symbol:
CTNND1
TMX2-CTNND1
Gene Name:
catenin delta 1
TMX2-CTNND1 readthrough (NMD candidate)
Gene Aliases:
CAS, CTNND, P120CAS, P120CTN, p120, p120(CAS), p120(CTN)
-
Chromosome Location:
Chr.11: 57761762 - 57819180 on Build GRCh38
Chromosome Location:
Chr.11: 57712523 - 57819180 on Build GRCh38
UniGene ID:
Hs.166011
Hs.166011
Species:
Human
Species Specific ID (Flybase ID):
-
-
siRNA ID
HSS102464
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CTNND1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001085458.1
NP_001078927.1
10
2388
NM_001085459.1
NP_001078928.1
10
2388
NM_001085460.1
NP_001078929.1
10
2315
NM_001085461.1
NP_001078930.1
10
2388
NM_001085462.1
NP_001078931.1
9
2196
NM_001085463.1
NP_001078932.1
8
1907
NM_001085464.1
NP_001078933.1
8
1907
NM_001085465.1
NP_001078934.1
8
1907
NM_001085466.1
NP_001078935.1
8
1907
NM_001085467.1
NP_001078936.1
8
1980
NM_001085468.1
NP_001078937.1
8
1907
NM_001085469.1
NP_001078938.1
7
1835
NM_001206883.1
NP_001193812.1
9
2087
NM_001206884.1
NP_001193813.1
9
2087
NM_001206885.1
NP_001193814.1
10
2388
NM_001206886.1
NP_001193815.1
9
2087
NM_001206887.1
NP_001193816.1
9
2087
NM_001206888.1
NP_001193817.1
9
2087
NM_001206889.1
NP_001193818.1
10
2206
NM_001206890.1
NP_001193819.1
8
1929
NM_001206891.1
NP_001193820.1
10
2279
NM_001331.2
NP_001322.1
10
2388
GenBank mRNA
AF062317.1
-
10
2353
AF062318.1
-
5
1111
AF062319.1
-
9
2052
AF062320.1
-
9
2052
AF062321.1
-
10
2353
AF062322.1
-
9
2052
AF062323.1
-
8
1872
AF062324.1
-
10
2280
AF062325.1
-
9
2052
AF062326.1
-
5
1111
AF062327.1
-
9
2052
AF062328.1
-
10
2353
AF062329.1
-
10
2171
AF062330.1
-
8
1894
AF062331.1
-
10
2244
AF062332.1
-
8
1872
AF062333.1
-
5
1111
AF062334.1
-
8
1945
AF062335.1
-
8
1872
AF062336.1
-
7
1800
AF062337.1
-
8
1872
AF062338.1
-
8
1872
AF062339.1
-
10
2353
AF062340.1
-
5
1184
AF062341.1
-
10
2353
AF062342.1
-
5
1111
AF062343.1
-
9
2161
AF062344.1
-
5
1111
AK292554.1
-
10
2330
BC010501.1
-
6
1509
BC075795.1
-
10
2204
5269008
BT007333.1
-
4
848
CR749275.1
-
10
2148
Gene Symbol: TMX2-CTNND1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NR_037646.1
11
2376
GenBank mRNA
AB002382.1
-
11
2282
Back To Top
More Information
Panther Classification:
Molecular Function -
intermediate filament binding protein
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
cell adhesion
brain development
Wnt signaling pathway
single organismal cell-cell adhesion
adherens junction organization
negative regulation of canonical Wnt signaling pathway
receptor binding
protein binding
protein kinase binding
cadherin binding
cadherin binding involved in cell-cell adhesion
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0