Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other DNM2 SIRNA Assays ›
          Gene Symbol:
          DNM2
          Gene Name:
          dynamin 2
          Gene Aliases:
          CMT2M, CMTDI1, CMTDIB, DI-CMTB, DYN2, DYNII, LCCS5
          Chromosome Location:
          Chr.19: 10718053 - 10831910 on Build GRCh38
          UniGene ID:
          Hs.211463
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS102857
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: DNM2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001005360.2 NP_001005360.1 3 477
          NM_001005361.2 NP_001005361.1 3 477
          NM_001005362.2 NP_001005362.1 3 477
          NM_001190716.1 NP_001177645.1 3 477
          NM_004945.3 NP_004936.2 3 477
          GenBank mRNA AB209213.1 - 4 384
          AK289831.1 - 3 322
          AK312260.1 - 3 451
          BC039596.1 - 3 367 5722134
          BC054501.1 - 3 437 6473791
          L36983.1 - 3 431

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_013199, XM_006242614, XM_006242611, XM_006242613, XM_008765917, and others.
          This siRNA also targets the following transcripts in Mouse: NM_001253894, NM_001253893, NM_007871, NM_001039520, L31398, and others.

          Panther Classification:

          Molecular Function -

          membrane traffic protein

          Gene Ontology Categories:

          Function(s) Process(es)

          G2/M transition of mitotic cell cycle
          G-protein coupled receptor internalization
          ventricular septum development
          regulation of transcription, DNA-templated
          Golgi to plasma membrane transport
          endocytosis
          receptor-mediated endocytosis
          phagocytosis
          signal transduction
          spermatogenesis
          response to light stimulus
          positive regulation of lamellipodium assembly
          antigen processing and presentation of exogenous peptide antigen via MHC class II
          negative regulation of transforming growth factor beta receptor signaling pathway
          regulation of axon extension
          receptor internalization
          transferrin transport
          regulation of Rac protein signal transduction
          aorta development
          response to cocaine
          positive regulation of apoptotic process
          macropinocytosis
          positive regulation of nitric oxide biosynthetic process
          positive regulation of transcription, DNA-templated
          synaptic vesicle transport
          neuron projection morphogenesis
          positive regulation of phagocytosis
          regulation of nitric-oxide synthase activity
          cilium morphogenesis
          coronary vasculature development
          cellular response to carbon monoxide
          cellular response to X-ray
          cellular response to nitric oxide
          positive regulation of substrate adhesion-dependent cell spreading
          cellular response to dopamine
          regulation of Golgi organization
          positive regulation of sodium:potassium-exchanging ATPase activity
          negative regulation of membrane tubulation
          GTPase activity
          protein binding
          GTP binding
          microtubule binding
          SH3 domain binding
          enzyme binding
          protein kinase binding
          D2 dopamine receptor binding
          protein complex binding
          phosphatidylinositol 3-kinase regulatory subunit binding
          WW domain binding
          nitric-oxide synthase binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0