Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other EDNRA SIRNA Assays ›
          Gene Symbol:
          EDNRA
          Gene Name:
          endothelin receptor type A
          Gene Aliases:
          ET-A, ETA, ETA-R, ETAR, ETRA, MFDA, hET-AR
          Chromosome Location:
          Chr.4: 147480917 - 147544954 on Build GRCh38
          UniGene ID:
          Hs.183713
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS103071
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: EDNRA
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001256283.1 NP_001243212.1 2 497
          NM_001957.3 NP_001948.1 3 987
          NR_045958.1 3 987
          GenBank mRNA AK304451.1 - 2 269
          AK312812.1 - 3 807
          AK315931.1 - 2 482
          AY275462.1 - 2 457
          BC022511.1 - 3 807 4812050
          D90348.1 - 3 937
          L06622.1 - 2 463
          S45956.1 - 3 678
          S57498.1 - 3 941
          S63938.1 - 2 530
          S67127.1 - 2 594
          S81539.1 - 2 457
          S81542.1 - 2 457
          X61950.1 - 3 941

          Back To Top

          More Information


          Set Membership:

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          patterning of blood vessels
          response to hypoxia
          in utero embryonic development
          histamine secretion
          glomerular filtration
          smooth muscle contraction
          signal transduction
          G-protein coupled receptor signaling pathway
          activation of adenylate cyclase activity
          activation of phospholipase C activity
          positive regulation of cytosolic calcium ion concentration
          protein kinase C-activating G-protein coupled receptor signaling pathway
          Rho protein signal transduction
          heart development
          aging
          respiratory gaseous exchange
          regulation of blood pressure
          cell proliferation
          positive regulation of cell proliferation
          neural crest cell development
          artery smooth muscle contraction
          glucose transport
          sensory perception of pain
          negative regulation of cAMP biosynthetic process
          response to lipopolysaccharide
          vasoconstriction
          positive regulation of odontogenesis
          negative regulation of apoptotic process
          penile erection
          response to morphine
          fibroblast proliferation
          enteric nervous system development
          smooth muscle cell proliferation
          regulation of epithelial cell proliferation
          positive regulation of inflammatory response
          positive regulation of release of sequestered calcium ion into cytosol
          positive regulation of cytosolic calcium ion concentration involved in phospholipase C-activating G-protein coupled signaling pathway
          maternal process involved in parturition
          head development
          positive regulation of ERK1 and ERK2 cascade
          cellular response to mechanical stimulus
          endothelin receptor signaling pathway
          positive regulation of neutrophil chemotaxis
          positive regulation of kidney development
          phosphatidylinositol phospholipase C activity
          endothelin receptor activity
          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-st8h5:80/100.66.79.144:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0