Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ELF2 SIRNA Assays ›
Gene Symbol:
ELF2
Gene Name:
E74 like ETS transcription factor 2
Gene Aliases:
EU32, NERF, NERF-1A, NERF-1B, NERF-1a,b, NERF-2
Chromosome Location:
Chr.4: 139057717 - 139177218 on Build GRCh38
UniGene ID:
Hs.634040
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS103184
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ELF2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001276457.1
NP_001263386.1
6
1674
NM_001276458.1
NP_001263387.1
7
1725
NM_001276459.1
NP_001263388.1
6
1638
NM_006874.3
NP_006865.1
7
1761
NM_201999.2
NP_973728.1
9
1911
XM_005262803.2
XP_005262860.1
10
2220
XM_005262804.2
XP_005262861.1
10
2184
XM_005262805.2
XP_005262862.1
9
2097
XM_006714128.2
XP_006714191.1
9
2133
XM_006714129.3
XP_006714192.1
9
1999
XM_011531710.2
XP_011530012.1
10
2177
XM_011531711.2
XP_011530013.1
8
1726
XM_011531712.2
XP_011530014.1
7
1717
XM_011531714.2
XP_011530016.1
4
1194
XM_017007856.1
XP_016863345.1
9
2019
XM_017007857.1
XP_016863346.1
9
1963
XM_017007858.1
XP_016863347.1
8
1968
XM_017007859.1
XP_016863348.1
9
2054
XM_017007860.1
XP_016863349.1
8
1876
XM_017007861.1
XP_016863350.1
9
2004
GenBank mRNA
AF256222.1
-
9
1863
AF256223.1
-
7
1614
AK301307.1
-
7
1606
AK303782.1
-
4
1464
AK316455.1
-
7
1636
BC034951.1
-
7
1845
4821608
BC065025.1
-
6
1604
6044351
U43188.1
-
9
1887
U43189.1
-
7
1658
Back To Top
More Information
Panther Classification:
Molecular Function -
winged helix/forkhead transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
cell differentiation
positive regulation of transcription, DNA-templated
RNA polymerase II transcription factor activity, sequence-specific DNA binding
protein binding
sequence-specific DNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-rhr4g:80/100.66.78.163:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0