Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MARK2 SIRNA Assays ›
          Gene Symbol:
          MARK2
          Gene Name:
          microtubule affinity regulating kinase 2
          Gene Aliases:
          EMK-1, EMK1, PAR-1, Par-1b, Par1b
          Chromosome Location:
          Chr.11: 63838928 - 63911020 on Build GRCh38
          UniGene ID:
          Hs.567261
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS103218
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: MARK2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001039469.2 NP_001034558.2 8 1128
          NM_001163296.1 NP_001156768.1 8 1128
          NM_001163297.1 NP_001156769.1 8 1128
          NM_004954.4 NP_004945.4 8 1128
          NM_017490.3 NP_059672.2 8 945
          XM_011544789.1 XP_011543091.1 9 1019
          XM_011544790.1 XP_011543092.1 9 1019
          XM_011544791.1 XP_011543093.1 9 1019
          XM_011544792.1 XP_011543094.1 9 1019
          XM_011544793.1 XP_011543095.1 9 1019
          XM_011544794.1 XP_011543096.1 9 1019
          XM_011544795.1 XP_011543097.1 9 1019
          XM_011544796.1 XP_011543098.1 9 1019
          XM_011544797.1 XP_011543099.1 9 1019
          XM_011544798.1 XP_011543100.1 9 1019
          XM_017017310.1 XP_016872799.1 8 1101
          XM_017017311.1 XP_016872800.1 9 1019
          GenBank mRNA AB188493.1 - 8 1132
          AB271694.1 - 7 450
          AF387638.1 - 7 468
          AK225766.1 - 8 963
          AK290339.1 - 8 913
          AK307858.1 - 9 764
          BC008771.2 - 8 520
          BC084540.1 - 8 587 4591688
          BT007342.1 - 7 450
          X97630.1 - 8 857

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          microtubule cytoskeleton organization
          mitophagy
          neuron migration
          protein phosphorylation
          positive regulation of neuron projection development
          Wnt signaling pathway
          peptidyl-threonine phosphorylation
          establishment of cell polarity
          activation of protein kinase activity
          intracellular signal transduction
          establishment or maintenance of epithelial cell apical/basal polarity
          protein autophosphorylation
          regulation of axonogenesis
          regulation of cytoskeleton organization
          mitochondrion localization
          cell-cell adhesion
          magnesium ion binding
          protein serine/threonine kinase activity
          protein binding
          ATP binding
          lipid binding
          protein kinase activator activity
          poly(A) RNA binding
          tau-protein kinase activity
          cadherin binding involved in cell-cell adhesion

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0