Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ERCC3 SIRNA Assays ›
          Gene Symbol:
          ERCC3
          Gene Name:
          ERCC excision repair 3, TFIIH core complex helicase subunit
          Gene Aliases:
          BTF2, GTF2H, RAD25, TFIIH, TTD2, XPB
          Chromosome Location:
          Chr.2: 127257290 - 127294176 on Build GRCh38
          UniGene ID:
          Hs.469872
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS103349
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ERCC3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000122.1 NP_000113.1 13 2136
          NM_001303416.1 NP_001290345.1 13 2251
          NM_001303418.1 NP_001290347.1 13 2313
          XM_011510794.1 XP_011509096.1 13 2115
          XM_011510795.1 XP_011509097.1 13 1989
          XM_017003583.1 XP_016859072.1 13 1971
          GenBank mRNA AK091500.1 - 13 2238
          AK095557.1 - 13 2313
          AK126697.1 - 1 2193
          AK127469.1 - 9 4012
          AK290474.1 - 13 2062
          BC008820.2 - 13 2105 3958781
          M31899.1 - 13 2136

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          DNA metabolism protein DNA helicase nucleic acid metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          nucleotide-excision repair, DNA duplex unwinding
          response to hypoxia
          DNA topological change
          DNA repair
          transcription-coupled nucleotide-excision repair
          nucleotide-excision repair
          nucleotide-excision repair, preincision complex stabilization
          nucleotide-excision repair, preincision complex assembly
          nucleotide-excision repair, DNA incision, 3'-to lesion
          nucleotide-excision repair, DNA incision, 5'-to lesion
          transcription initiation from RNA polymerase I promoter
          transcription elongation from RNA polymerase I promoter
          termination of RNA polymerase I transcription
          transcription from RNA polymerase II promoter
          transcription initiation from RNA polymerase II promoter
          transcription elongation from RNA polymerase II promoter
          7-methylguanosine mRNA capping
          protein phosphorylation
          apoptotic process
          response to oxidative stress
          protein localization
          response to UV
          UV protection
          viral process
          nucleotide-excision repair, DNA incision
          hair cell differentiation
          positive regulation of apoptotic process
          positive regulation of transcription from RNA polymerase II promoter
          embryonic organ development
          global genome nucleotide-excision repair
          regulation of mitotic cell cycle phase transition
          DNA binding
          damaged DNA binding
          ATP-dependent DNA helicase activity
          helicase activity
          protein kinase activity
          protein binding
          ATP binding
          GTP binding
          protein C-terminus binding
          DNA-dependent ATPase activity
          transcription factor binding
          RNA polymerase II carboxy-terminal domain kinase activity
          ATPase activity
          dATP binding
          peptide binding
          3'-5' DNA helicase activity
          protein N-terminus binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0