Search
Search

EXTL1| Gene Symbol: EXTL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004455.2 | NP_004446.2 | 1 | 1154 | ||
| XM_005245779.3 | XP_005245836.1 | 1 | 1185 | |||
| XM_017000650.1 | XP_016856139.1 | 1 | 423 | |||
| GenBank mRNA | BC065528.1 | - | 1 | 1000 | 6150218 | |
| U67191.1 | - | 1 | 1153 | |||