Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other F2RL1 SIRNA Assays ›
          Gene Symbol:
          F2RL1
          Gene Name:
          F2R like trypsin receptor 1
          Gene Aliases:
          GPR11, PAR2
          Chromosome Location:
          Chr.5: 76819008 - 76835315 on Build GRCh38
          UniGene ID:
          Hs.744181
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS103473
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: F2RL1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_017009223.1 XP_016864712.1 2 539
          GenBank mRNA AY336105.1 - 2 561
          BC012453.1 - 2 572 3857382
          BC018130.1 - 2 634 3895653
          BT009856.1 - 2 480
          U34038.1 - 2 627

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          T cell activation involved in immune response
          positive regulation of leukocyte chemotaxis
          positive regulation of cytokine secretion involved in immune response
          positive regulation of glomerular filtration
          inflammatory response
          G-protein coupled receptor signaling pathway
          positive regulation of cytosolic calcium ion concentration
          blood coagulation
          negative regulation of tumor necrosis factor-mediated signaling pathway
          positive regulation of phosphatidylinositol 3-kinase signaling
          regulation of blood coagulation
          positive regulation of cell migration
          positive regulation of actin filament depolymerization
          positive regulation of pseudopodium assembly
          positive regulation of superoxide anion generation
          positive regulation of toll-like receptor 2 signaling pathway
          negative regulation of toll-like receptor 3 signaling pathway
          positive regulation of toll-like receptor 3 signaling pathway
          positive regulation of toll-like receptor 4 signaling pathway
          positive regulation of Rho protein signal transduction
          chemokine (C-C motif) ligand 2 secretion
          neutrophil activation
          regulation of I-kappaB kinase/NF-kappaB signaling
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          positive regulation of eosinophil degranulation
          innate immune response
          positive regulation of vasodilation
          positive regulation of transcription from RNA polymerase II promoter
          regulation of JNK cascade
          negative regulation of JNK cascade
          positive regulation of JNK cascade
          interleukin-1 beta secretion
          leukocyte migration
          positive regulation of chemotaxis
          positive regulation of positive chemotaxis
          defense response to virus
          positive regulation of phagocytosis, engulfment
          establishment of endothelial barrier
          positive regulation of ERK1 and ERK2 cascade
          thrombin receptor signaling pathway
          leukocyte proliferation
          positive regulation of neutrophil mediated killing of gram-negative bacterium
          interleukin-10 secretion
          interferon-gamma secretion
          chemokine secretion
          negative regulation of chemokine secretion
          mature conventional dendritic cell differentiation
          positive regulation of renin secretion into blood stream
          positive regulation of interleukin-8 secretion
          positive regulation of interleukin-6 secretion
          G-protein alpha-subunit binding
          receptor activity
          G-protein coupled receptor activity
          receptor binding
          protein binding
          thrombin receptor activity
          G-protein beta-subunit binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0