Search
Search

GLI1| Gene Symbol: GLI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001160045.1 | NP_001153517.1 | 10 | 1639 | ||
| NM_001167609.1 | NP_001161081.1 | 11 | 1720 | |||
| NM_005269.2 | NP_005260.1 | 12 | 1843 | |||
| XM_011538189.2 | XP_011536491.1 | 12 | 2089 | |||
| XM_011538190.1 | XP_011536492.1 | 11 | 1625 | |||
| GenBank mRNA | BC013000.2 | - | 11 | 1803 | 3531657 | |
| GQ890670.2 | - | 10 | 1642 | |||
| X07384.1 | - | 12 | 1843 | |||