Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other GSN SIRNA Assays ›
Gene Symbol:
GSN
Gene Name:
gelsolin
Gene Aliases:
ADF, AGEL
Chromosome Location:
Chr.9: 121201082 - 121332844 on Build GRCh38
UniGene ID:
Hs.522373
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS104524
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: GSN
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000177.4
NP_000168.1
9
1230
NM_001127662.1
NP_001121134.1
9
1143
NM_001127663.1
NP_001121135.2
11
1348
NM_001127664.1
NP_001121136.1
11
1315
NM_001127665.1
NP_001121137.1
11
1310
NM_001127666.1
NP_001121138.1
11
1307
NM_001127667.1
NP_001121139.1
11
1298
NM_001258029.1
NP_001244958.1
9
1130
NM_001258030.1
NP_001244959.1
9
1207
NM_198252.2
NP_937895.1
10
1236
XM_005251943.1
XP_005252000.1
10
1334
XM_005251944.1
XP_005252001.1
10
1325
XM_005251945.3
XP_005252002.1
10
1238
XM_006717079.1
XP_006717142.1
10
1172
XM_011518584.1
XP_011516886.1
11
1325
XM_011518585.1
XP_011516887.1
9
1145
XM_011518586.1
XP_011516888.1
12
1348
XM_011518587.2
XP_011516889.1
12
1390
XM_011518588.2
XP_011516890.1
12
1352
XM_011518589.2
XP_011516891.1
12
1348
XM_011518590.2
XP_011516892.1
12
1300
XM_011518591.2
XP_011516893.1
17
2392
XM_011518592.1
XP_011516894.1
11
1214
XM_011518593.1
XP_011516895.1
12
1360
XM_011518594.1
XP_011516896.1
6
557
XM_017014643.1
XP_016870132.1
18
2543
XM_017014644.1
XP_016870133.1
18
2477
XM_017014645.1
XP_016870134.1
10
1289
XM_017014646.1
XP_016870135.1
13
1460
XM_017014647.1
XP_016870136.1
19
2676
XM_017014648.1
XP_016870137.1
12
1287
GenBank mRNA
AK092797.1
-
5
1371
AK096280.1
-
10
1209
AK125819.1
-
18
2039
AK295201.1
-
10
1074
AK295572.1
-
11
1269
AK297870.1
-
9
1088
AK299453.1
-
9
1195
AK300631.1
-
11
1282
AK304629.1
-
9
1151
AK304675.1
-
8
1184
AK307859.1
-
9
1094
AK315494.1
-
9
1201
BC026033.1
-
9
1174
4661084
BX647999.1
-
9
1132
X04412.1
-
9
1183
Back To Top
More Information
Panther Classification:
Molecular Function -
non-motor actin binding protein
Gene Ontology Categories:
Function(s)
Process(es)
phagocytosis, engulfment
aging
positive regulation of gene expression
oligodendrocyte development
striated muscle atrophy
extracellular matrix disassembly
actin filament polymerization
protein destabilization
tissue regeneration
sequestering of actin monomers
cellular protein metabolic process
actin nucleation
response to ethanol
negative regulation of viral entry into host cell
phosphatidylinositol-mediated signaling
actin filament severing
barbed-end actin filament capping
positive regulation of actin nucleation
response to folic acid
actin filament capping
cilium morphogenesis
cellular response to cadmium ion
regulation of podosome assembly
actin filament reorganization
renal protein absorption
hepatocyte apoptotic process
positive regulation of keratinocyte apoptotic process
regulation of wound healing, spreading of epidermal cells
regulation of establishment of T cell polarity
regulation of plasma membrane raft polarization
regulation of receptor clustering
positive regulation of protein processing in phagocytic vesicle
amyloid fibril formation
positive regulation of cysteine-type endopeptidase activity involved in apoptotic signaling pathway
actin binding
calcium ion binding
protein binding
protein domain specific binding
myosin II binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0