Search
Search

HOXD8| Gene Symbol: HOXD8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199746.1 | NP_001186675.1 | 2 | 1481 | ||
| NM_001199747.1 | NP_001186676.1 | 3 | 509 | |||
| NM_019558.3 | NP_062458.1 | 2 | 1484 | |||
| GenBank mRNA | BC038709.1 | - | 2 | 1331 | 4614350 | |
| BC090853.1 | - | 3 | 1180 | 6180525 | ||
| BI764317.1 | - | 2 | 308 | |||
| BQ429720.1 | - | 3 | 459 | |||