Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other IKBKB SIRNA Assays ›
Gene Symbol:
IKBKB
Gene Name:
inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase beta
Gene Aliases:
IKK-beta, IKK2, IKKB, IMD15, NFKBIKB
Chromosome Location:
Chr.8: 42271302 - 42332653 on Build GRCh38
UniGene ID:
Hs.597664
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS105292
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Contact Us
Your Price
Online offer:
3062.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: IKBKB
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001190720.2
NP_001177649.1
12, 13
1431
NM_001242778.1
NP_001229707.1
12, 13
1459
NM_001556.2
NP_001547.1
13, 14
1554
NR_033818.1
13, 14
1610
NR_033819.1
13, 14
1554
NR_040009.1
13, 14
1501
XM_005273490.2
XP_005273547.1
13, 14
1554
XM_005273491.4
XP_005273548.1
12, 13
1454
XM_005273492.3
XP_005273549.1
13, 14
1554
XM_005273493.3
XP_005273550.1
10, 11
1250
XM_005273494.2
XP_005273551.1
13, 14
1554
XM_005273495.2
XP_005273552.1
12, 13
1295
XM_005273496.3
XP_005273553.1
11, 12
1315
XM_005273498.3
XP_005273555.1
9, 10
1124
XM_011544517.2
XP_011542819.1
13, 14
1552
XM_011544518.2
XP_011542820.1
12, 13
1404
XM_011544519.2
XP_011542821.1
12, 13
1422
XM_011544520.2
XP_011542822.1
10, 11
1269
XM_011544521.2
XP_011542823.1
11, 12
1382
XM_011544522.2
XP_011542824.1
10, 11
1230
XM_017013396.1
XP_016868885.1
12, 13
1507
GenBank mRNA
AB209090.1
-
12, 13
1372
AF029684.1
-
12, 13
1368
AF031416.1
-
12, 13
1368
AF080158.1
-
13, 14
1511
AK091297.1
-
7, 8
950
AK290952.1
-
13, 14
1420
AK298241.1
-
10, 11
1118
AK302723.1
-
12, 13
1415
AK303294.1
-
12, 13
1443
AK303528.1
-
12, 13
1447
AK316418.1
-
12, 13
1415
BC144080.1
-
12, 13
1345
BX648165.1
-
12, 13
2006
Back To Top
More Information
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
protein modifying enzyme
Gene Ontology Categories:
Function(s)
Process(es)
stimulatory C-type lectin receptor signaling pathway
skeletal muscle contraction
protein phosphorylation
inflammatory response
I-kappaB kinase/NF-kappaB signaling
I-kappaB phosphorylation
positive regulation of cell proliferation
response to virus
response to toxic substance
regulation of tumor necrosis factor-mediated signaling pathway
positive regulation of neuron projection development
cortical actin cytoskeleton organization
neuron projection development
response to lipopolysaccharide
tumor necrosis factor-mediated signaling pathway
negative regulation of myosin-light-chain-phosphatase activity
TRIF-dependent toll-like receptor signaling pathway
Fc-epsilon receptor signaling pathway
regulation of phosphorylation
response to drug
serine phosphorylation of STAT protein
negative regulation of apoptotic process
positive regulation of I-kappaB kinase/NF-kappaB signaling
innate immune response
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
T cell receptor signaling pathway
positive regulation of NF-kappaB transcription factor activity
stress-activated MAPK cascade
nucleotide-binding oligomerization domain containing signaling pathway
interleukin-1-mediated signaling pathway
response to fatty acid
cellular response to tumor necrosis factor
establishment of protein localization to plasma membrane
positive regulation of neuron death
regulation of establishment of endothelial barrier
negative regulation of bicellular tight junction assembly
protein kinase activity
protein serine/threonine kinase activity
protein binding
ATP binding
IkappaB kinase activity
protein kinase binding
protein homodimerization activity
protein heterodimerization activity
scaffold protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-nwsk8:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0