Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other IRF3 SIRNA Assays ›
          Gene Symbol:
          IRF3
          Gene Name:
          interferon regulatory factor 3
          Gene Aliases:
          IIAE7
          Chromosome Location:
          Chr.19: 49659569 - 49665875 on Build GRCh38
          UniGene ID:
          Hs.289052
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS105505
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: IRF3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001197122.1 NP_001184051.1 7, 8 1356
          NM_001197123.1 NP_001184052.1 7, 8 1398
          NM_001197124.1 NP_001184053.1 6, 7 959
          NM_001197125.1 NP_001184054.1 6, 7 1167
          NM_001197126.1 NP_001184055.1 5, 6 995
          NM_001197127.1 NP_001184056.1 5, 6 786
          NM_001197128.1 NP_001184057.1 4, 5 614
          NM_001571.5 NP_001562.1 7, 8 1340
          NR_045568.1 7, 8 1487
          XM_006723197.1 XP_006723260.1 7, 8 1334
          XM_006723198.1 XP_006723261.1 7, 8 1270
          XM_006723200.1 XP_006723263.1 6, 7 1019
          XM_006723201.1 XP_006723264.1 6, 7 1092
          XM_006723202.2 XP_006723265.1 7, 8 1241
          XM_017026766.1 XP_016882255.1 7, 8 1309
          XM_017026767.1 XP_016882256.1 7, 8 1254
          GenBank mRNA AA568913.1 - 1, 2 278
          AB102886.1 - 2, 3 268
          AB102887.1 - 5, 6 706
          AK292027.1 - 7, 8 1469
          AK314421.1 - 7, 8 1253
          BC000660.2 - 7, 8 1258 3347928
          BC009395.2 - 7, 8 1283 4135383
          BC071721.1 - 6, 7 1429 5494536
          BM477213.1 - 2, 3 218
          BU859806.1 - 2, 3 491
          HM357932.1 - 7, 8 1228
          HM357933.1 - 4, 5 444
          HM357934.1 - 5, 6 825
          HM357935.1 - 5, 6 616
          HM357936.1 - 6, 7 789
          HM357937.1 - 6, 7 997
          Z56281.1 - 7, 8 1133

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          winged helix/forkhead transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          transcription from RNA polymerase II promoter
          apoptotic process
          cellular response to DNA damage stimulus
          lipopolysaccharide-mediated signaling pathway
          negative regulation of type I interferon production
          positive regulation of type I interferon production
          positive regulation of interferon-alpha production
          positive regulation of interferon-beta production
          TRIF-dependent toll-like receptor signaling pathway
          MDA-5 signaling pathway
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          response to exogenous dsRNA
          type I interferon biosynthetic process
          positive regulation of cytokine secretion
          defense response to virus
          interferon-gamma-mediated signaling pathway
          type I interferon signaling pathway
          positive regulation of type I interferon-mediated signaling pathway
          cellular response to dsRNA
          macrophage apoptotic process
          programmed necrotic cell death
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
          DNA binding
          transcription cofactor activity
          protein binding
          identical protein binding
          protein homodimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0