Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other JUP SIRNA Assays ›
          Gene Symbol:
          JUP
          Gene Name:
          junction plakoglobin
          Gene Aliases:
          ARVD12, CTNNG, DP3, DPIII, PDGB, PKGB
          Chromosome Location:
          Chr.17: 41754607 - 41786853 on Build GRCh38
          UniGene ID:
          Hs.514174
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS105646
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: JUP
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_002230.2 NP_002221.1 6 1084
          NM_021991.2 NP_068831.1 6 1084
          XM_006721873.2 XP_006721936.1 7 1334
          XM_006721874.2 XP_006721937.1 6 1247
          XM_006721875.1 XP_006721938.1 6 1166
          XM_006721878.1 XP_006721941.1 6 1147
          XM_011524753.2 XP_011523055.1 7 1370
          XM_011524755.1 XP_011523057.1 6 1079
          XM_011524756.1 XP_011523058.1 6 977
          XM_011524757.2 XP_011523059.1 6 1244
          XM_011524758.1 XP_011523060.1 6 973
          XM_017024588.1 XP_016880077.1 6 1066
          XM_017024589.1 XP_016880078.1 6 1067
          XM_017024590.1 XP_016880079.1 6 973
          GenBank mRNA AK226051.1 - 6 1061
          AY243535.1 - 5 957
          BC000441.2 - 6 1070
          BC011865.2 - 6 1228 4542042
          KU178155.1 - 5 951
          M23410.1 - 6 1067
          Z68228.1 - 6 1070

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          desmosome assembly
          cytoskeletal anchoring at plasma membrane
          signal transduction
          single organismal cell-cell adhesion
          cell migration
          adherens junction organization
          adherens junction assembly
          regulation of cell proliferation
          positive regulation of protein import into nucleus
          skin development
          detection of mechanical stimulus
          positive regulation of sequence-specific DNA binding transcription factor activity
          protein heterooligomerization
          endothelial cell-cell adhesion
          cellular response to indole-3-methanol
          bundle of His cell-Purkinje myocyte adhesion involved in cell communication
          regulation of heart rate by cardiac conduction
          establishment of protein localization to plasma membrane
          positive regulation of canonical Wnt signaling pathway
          regulation of ventricular cardiac muscle cell action potential
          transcription coactivator activity
          signal transducer activity
          structural molecule activity
          structural constituent of cell wall
          protein binding
          protein kinase binding
          protein phosphatase binding
          protein homodimerization activity
          alpha-catenin binding
          cadherin binding
          cell adhesion molecule binding
          cell adhesive protein binding involved in bundle of His cell-Purkinje myocyte communication
          cadherin binding involved in cell-cell adhesion

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0