Search
Search

KCNA7| Gene Symbol: KCNA7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031886.2 | NP_114092.2 | 2 | 1244 | ||
| GenBank mRNA | AF315818.1 | - | 2 | 925 | ||
| AJ310479.1 | - | 2 | 1244 | |||
| AY779768.1 | - | 2 | 917 | |||
| BC140906.1 | - | 2 | 1038 | 9056718 | ||