Search
Search

LSP1| Gene Symbol: LSP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013253.1 | NP_001013271.1 | 5, 6 | 851 | ||
| NM_001013254.1 | NP_001013272.1 | 5, 6 | 1081 | |||
| NM_001013255.1 | NP_001013273.1 | 5, 6 | 587 | |||
| NM_001242932.1 | NP_001229861.1 | 6, 7 | 1082 | |||
| NM_001289005.1 | NP_001275934.1 | 5, 6 | 947 | |||
| NM_002339.2 | NP_002330.1 | 5, 6 | 765 | |||
| GenBank mRNA | AK056576.1 | - | 5, 6 | 1706 | ||
| AK092071.1 | - | 6, 7 | 1082 | |||
| AK093859.1 | - | 4, 5 | 1088 | |||
| AK222733.1 | - | 5, 6 | 661 | |||
| AK290279.1 | - | 5, 6 | 603 | |||
| AK292033.1 | - | 5, 6 | 949 | |||
| BC001785.1 | - | 5, 6 | 657 | 3542307 | ||
| BQ893037.1 | - | 5, 6 | 625 | |||
| CR541728.1 | - | 5, 6 | 590 | |||
| CR541754.1 | - | 5, 6 | 590 | |||
| S67783.1 | - | 5, 6 | 587 | |||
| X55188.1 | - | 5, 6 | 637 | |||