Search
Search

MAGEA8| Gene Symbol: MAGEA8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166400.1 | NP_001159872.1 | 4 | 924 | ||
| NM_001166401.1 | NP_001159873.1 | 3 | 896 | |||
| NM_005364.4 | NP_005355.2 | 3 | 645 | |||
| XM_017029520.1 | XP_016885009.1 | 4 | 712 | |||
| GenBank mRNA | AK313943.1 | - | 3 | 645 | ||
| BC002455.2 | - | 3 | 591 | 3345962 | ||
| BC012744.2 | - | 3 | 584 | 3617812 | ||
| BC098120.1 | - | 3 | 520 | |||
| BT007340.1 | - | 1 | 375 | |||