Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MEF2A SIRNA Assays ›
          Gene Symbol:
          MEF2A
          Gene Name:
          myocyte enhancer factor 2A
          Gene Aliases:
          ADCAD1, RSRFC4, RSRFC9, mef2
          Chromosome Location:
          Chr.15: 99565455 - 99716488 on Build GRCh38
          UniGene ID:
          Hs.268675
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS106435
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price 459.00
          Your Price
          Online offer:
          459.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: MEF2A
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001130926.2 NP_001124398.1 8, 9 1296
          NM_001130927.2 NP_001124399.1 8, 9 1116
          NM_001130928.2 NP_001124400.1 6, 7 915
          NM_001171894.2 NP_001165365.1 10, 11 1586
          NM_001319206.1 NP_001306135.1 10, 11 1402
          NM_005587.3 NP_005578.2 9, 10 1384
          XM_005254914.2 XP_005254971.1 9, 10 1361
          XM_005254915.2 XP_005254972.1 9, 10 1606
          XM_011521571.2 XP_011519873.1 10, 11 1385
          XM_011521572.2 XP_011519874.1 10, 11 1330
          XM_011521573.2 XP_011519875.1 10, 11 1360
          XM_011521576.2 XP_011519878.1 9, 10 1274
          XM_011521577.2 XP_011519879.1 9, 10 1208
          XM_011521578.2 XP_011519880.1 9, 10 1867
          XM_011521579.2 XP_011519881.1 9, 10 1284
          XM_011521581.2 XP_011519883.1 10, 11 1636
          XM_011521582.2 XP_011519884.1 11, 12 1841
          XM_011521583.2 XP_011519885.1 9, 10 1256
          XM_011521585.2 XP_011519887.1 9, 10 1444
          XM_011521586.2 XP_011519888.1 9, 10 1426
          XM_011521587.2 XP_011519889.1 8, 9 1402
          XM_017022190.1 XP_016877679.1 9, 10 1262
          XM_017022191.1 XP_016877680.1 9, 10 1254
          XM_017022192.1 XP_016877681.1 8, 9 1239
          XM_017022193.1 XP_016877682.1 9, 10 1422
          XM_017022194.1 XP_016877683.1 8, 9 1238
          XM_017022195.1 XP_016877684.1 8, 9 1095
          XM_017022196.1 XP_016877685.1 11, 12 1774
          XM_017022197.1 XP_016877686.1 10, 11 3553
          XM_017022198.1 XP_016877687.1 11, 12 1762
          XM_017022199.1 XP_016877688.1 8, 9 2039
          XM_017022200.1 XP_016877689.1 9, 10 1809
          XM_017022201.1 XP_016877690.1 8, 9 1432
          XM_017022202.1 XP_016877691.1 11, 12 1756
          GenBank mRNA AB208985.1 - 10, 11 1563
          AK291835.1 - 8, 9 1206
          AK294207.1 - 8, 9 1054
          AL831995.1 - 10, 11 1263
          BC013437.2 - 9, 10 1302 3896491
          BC053871.2 - 6, 7 887 5555678
          KU178181.1 - 7, 8 983
          KU178182.1 - 8, 9 1007
          KU178183.1 - 8, 9 1001
          KU178184.1 - 7, 8 977
          KU178185.1 - 6, 7 773
          X63381.1 - 8, 9 1211
          X68505.1 - 10, 11 1421
          Y16312.1 - 8, 9 1205

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          mitochondrial genome maintenance
          negative regulation of transcription from RNA polymerase II promoter
          MAPK cascade
          transcription, DNA-templated
          transcription from RNA polymerase II promoter
          apoptotic process
          heart development
          muscle organ development
          positive regulation of cardiac muscle hypertrophy
          positive regulation of transcription from RNA polymerase II promoter
          mitochondrion distribution
          dendrite morphogenesis
          positive regulation of muscle cell differentiation
          ventricular cardiac myofibril assembly
          cardiac conduction
          ERK5 cascade
          cellular response to calcium ion
          RNA polymerase II regulatory region sequence-specific DNA binding
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          RNA polymerase II transcription factor activity, sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          RNA polymerase II transcription factor binding
          RNA polymerase II transcription coactivator activity
          chromatin binding
          transcription factor activity, sequence-specific DNA binding
          transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding
          protein binding
          protein kinase binding
          activating transcription factor binding
          histone acetyltransferase binding
          histone deacetylase binding
          sequence-specific DNA binding
          SMAD binding
          protein heterodimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-st8h5:80/100.66.79.144:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0