Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MUTYH SIRNA Assays ›
Gene Symbol:
MUTYH
Gene Name:
mutY DNA glycosylase
Gene Aliases:
MYH
Chromosome Location:
Chr.1: 45329242 - 45340925 on Build GRCh38
UniGene ID:
Hs.271353
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS106809
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Contact Us
Your Price
Online offer:
3062.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MUTYH
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001048171.1
NP_001041636.1
5
607
NM_001048172.1
NP_001041637.1
5
498
NM_001048173.1
NP_001041638.1
5
495
NM_001048174.1
NP_001041639.1
5
414
NM_001128425.1
NP_001121897.1
5
649
NM_001293190.1
NP_001280119.1
5
610
NM_001293191.1
NP_001280120.1
5
447
NM_001293192.1
NP_001280121.1
5
543
NM_001293195.1
NP_001280124.1
6
564
NM_001293196.1
NP_001280125.1
5
432
NM_012222.2
NP_036354.1
5
640
XM_011541497.2
XP_011539799.1
5
706
XM_011541498.1
XP_011539800.1
5
484
XM_011541499.1
XP_011539801.1
5
470
XM_011541500.2
XP_011539802.1
5
665
XM_011541501.2
XP_011539803.1
6
579
XM_011541502.2
XP_011539804.1
5
484
XM_011541503.2
XP_011539805.1
5
494
XM_011541504.2
XP_011539806.1
5
533
XM_017001331.1
XP_016856820.1
5
483
XM_017001332.1
XP_016856821.1
5
485
XM_017001333.1
XP_016856822.1
5
538
XM_017001334.1
XP_016856823.1
5
431
XM_017001335.1
XP_016856824.1
5
377
GenBank mRNA
AB032920.1
-
5
558
AB032921.1
-
5
528
AB032922.1
-
5
525
AB032923.1
-
5
461
AB032924.1
-
5
425
AB032925.1
-
5
392
AB032926.1
-
6
542
AB032927.1
-
5
463
AB032928.1
-
5
460
AB032929.1
-
5
396
AL560568.3
-
5
416
BC003178.1
-
5
545
2958002
BI818560.1
-
5
317
BM926983.1
-
5
607
KU178203.1
-
5
391
Back To Top
More Information
Panther Classification:
Molecular Function -
DNA glycosylase
Gene Ontology Categories:
Function(s)
Process(es)
DNA repair
mismatch repair
response to oxidative stress
depurination
DNA binding
protein binding
DNA N-glycosylase activity
MutLalpha complex binding
MutLbeta complex binding
MutSalpha complex binding
MutSbeta complex binding
metal ion binding
4 iron, 4 sulfur cluster binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-55658555d6-9rjdb:80/100.66.79.224:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline