Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MYH6 SIRNA Assays ›
Gene Symbol:
MYH6
Gene Name:
myosin heavy chain 6
Gene Aliases:
ASD3, CMD1EE, CMH14, MYHC, MYHCA, SSS3, alpha-MHC
Chromosome Location:
Chr.14: 23381990 - 23408277 on Build GRCh38
UniGene ID:
Hs.278432
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS106862
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MYH6
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_002471.3
NP_002462.2
24
3274
GenBank mRNA
BC132667.1
-
23
3233
40146689
D00943.1
-
22
3203
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
in utero embryonic development
regulation of the force of heart contraction
regulation of heart rate
protein dephosphorylation
muscle contraction
striated muscle contraction
adult heart development
visceral muscle development
regulation of heart contraction
regulation of blood pressure
muscle filament sliding
myofibril assembly
BMP signaling pathway
regulation of ATPase activity
sarcomere organization
ATP metabolic process
cardiac muscle fiber development
atrial cardiac muscle tissue morphogenesis
ventricular cardiac muscle tissue morphogenesis
cardiac muscle contraction
canonical Wnt signaling pathway
regulation of heart growth
microfilament motor activity
actin binding
calmodulin binding
ATP binding
ATPase activity
myosin phosphatase activity
protein kinase binding
actin-dependent ATPase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0