Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MYO5A SIRNA Assays ›
          Gene Symbol:
          MYO5A
          Gene Name:
          myosin VA
          Gene Aliases:
          GS1, MYH12, MYO5, MYR12
          Chromosome Location:
          Chr.15: 52307283 - 52529050 on Build GRCh38
          UniGene ID:
          Hs.21213
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS106896
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: MYO5A
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000259.3 NP_000250.3 30 4173
          NM_001142495.1 NP_001135967.1 30 4173
          XM_005254397.3 XP_005254454.1 30 4005
          XM_011521606.2 XP_011519908.2 30 4008
          XM_011521607.2 XP_011519909.2 29 3999
          XM_011521608.2 XP_011519910.2 30 4007
          XM_011521609.2 XP_011519911.2 30 4007
          XM_011521610.2 XP_011519912.2 29 3999
          XM_011521611.2 XP_011519913.2 30 4007
          XM_011521612.2 XP_011519914.2 29 3998
          XM_017022227.1 XP_016877716.1 29 3900
          GenBank mRNA AB209487.1 - 23 2786
          AB209505.1 - 23 2786
          AB290180.1 - 30 3932
          AF090424.1 - 3 308
          AF090425.1 - 2 299
          AF090426.1 - 3 308
          AF090427.1 - 3 308
          AF090428.1 - 3 308
          EU921839.1 - 2 200
          S74799.1 - 7 750
          U90942.1 - 30 4179
          Y07759.1 - 30 4179

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          actin binding motor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          exocytosis
          post-Golgi vesicle-mediated transport
          chemical synaptic transmission
          visual perception
          vesicle-mediated transport
          actin filament-based movement
          vesicle transport along actin filament
          insulin secretion
          melanocyte differentiation
          regulation of inositol 1,4,5-trisphosphate-sensitive calcium-release channel activity
          locomotion involved in locomotory behavior
          secretory granule localization
          melanosome transport
          cellular response to insulin stimulus
          odontogenesis
          myelination
          long-chain fatty acid biosynthetic process
          synapse organization
          endoplasmic reticulum localization
          protein localization to plasma membrane
          regulation of Golgi organization
          microfilament motor activity
          actin binding
          calcium ion binding
          calmodulin binding
          ATP binding
          Rab GTPase binding
          poly(A) RNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0