Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SEPT2 SIRNA Assays ›
          Gene Symbol:
          SEPT2
          Gene Name:
          septin 2
          Gene Aliases:
          DIFF6, NEDD-5, NEDD5, Pnutl3, hNedd5
          Chromosome Location:
          Chr.2: 241315187 - 241354027 on Build GRCh38
          UniGene ID:
          Hs.721234
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS107067
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SEPT2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001008491.2 NP_001008491.1 6 868
          NM_001008492.2 NP_001008492.1 6 550
          NM_001282972.1 NP_001269901.1 6 599
          NM_001282973.1 NP_001269902.1 9 823
          NM_001321029.1 NP_001307958.1 6 584
          NM_001321030.1 NP_001307959.1 6 582
          NM_001321031.1 NP_001307960.1 7 685
          NM_001321032.1 NP_001307961.1 5 465
          NM_001321033.1 NP_001307962.1 7 701
          NM_001321034.1 NP_001307963.1 7 700
          NM_001321035.1 NP_001307964.1 6 566
          NM_004404.4 NP_004395.1 5 465
          NM_006155.2 NP_006146.1 5 767
          XM_011511237.1 XP_011509539.1 5 431
          XM_011511238.1 XP_011509540.1 4 299
          XM_011511240.1 XP_011509542.1 4 308
          XM_017004189.1 XP_016859678.1 7 660
          XM_017004190.1 XP_016859679.1 7 595
          XM_017004191.1 XP_016859680.1 7 660
          XM_017004192.1 XP_016859681.1 7 660
          XM_017004193.1 XP_016859682.1 5 465
          XM_017004194.1 XP_016859683.1 7 615
          XM_017004195.1 XP_016859684.1 7 819
          XM_017004196.1 XP_016859685.1 7 701
          XM_017004197.1 XP_016859686.1 6 516
          XM_017004198.1 XP_016859687.1 6 578
          XM_017004199.1 XP_016859688.1 7 617
          XM_017004200.1 XP_016859689.1 6 580
          XM_017004201.1 XP_016859690.1 6 702
          XM_017004202.1 XP_016859691.1 5 684
          XM_017004203.1 XP_016859692.1 6 508
          XM_017004204.1 XP_016859693.1 5 423
          XM_017004205.1 XP_016859694.1 7 615
          XM_017004206.1 XP_016859695.1 6 516
          XM_017004207.1 XP_016859696.1 3 317
          XM_017004208.1 XP_016859697.1 6 527
          XM_017004209.1 XP_016859698.1 5 414
          XM_017004210.1 XP_016859699.1 5 458
          XM_017004211.1 XP_016859700.1 4 311
          XM_017004212.1 XP_016859701.1 4 563
          XM_017004213.1 XP_016859702.1 7 660
          GenBank mRNA AF038404.1 - 5 404
          AK294563.1 - 6 507
          BC014455.1 - 6 747 4906575
          BC033559.1 - 9 801 4821654
          BG721140.1 - 5 468
          BX648000.1 - 6 484
          D28540.1 - 4 388
          D63878.1 - 5 584
          DB091579.1 - 5 465

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          cytoskeletal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of L-glutamate transport
          mitotic nuclear division
          smoothened signaling pathway
          neuron projection development
          regulation of protein localization
          cilium assembly
          regulation of catalytic activity
          cell division
          cell-cell adhesion
          protein binding
          GTP binding
          enzyme regulator activity
          protein complex scaffold
          cadherin binding involved in cell-cell adhesion

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-5thf6:80/100.66.79.225:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0