Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NFATC4 SIRNA Assays ›
Gene Symbol:
NFATC4
Gene Name:
nuclear factor of activated T-cells 4
Gene Aliases:
NF-AT3, NF-ATC4, NFAT3
Chromosome Location:
Chr.14: 24366911 - 24379604 on Build GRCh38
UniGene ID:
Hs.77810
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS107121
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NFATC4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001136022.2
NP_001129494.1
10
2465
NM_001198965.1
NP_001185894.1
9
2453
NM_001198966.2
NP_001185895.1
9
2107
NM_001198967.2
NP_001185896.1
10
2465
NM_001288802.1
NP_001275731.1
9
2256
NM_001320043.1
NP_001306972.1
10
2465
NM_004554.4
NP_004545.2
9
2453
XM_011536797.2
XP_011535099.1
9
2288
XM_011536798.1
XP_011535100.1
9
2378
XM_011536799.2
XP_011535101.1
9
2290
GenBank mRNA
AK293185.1
-
9
2328
AK302271.1
-
9
2093
AK310433.1
-
9
2256
AK310445.1
-
10
2437
BC008857.2
-
8
1954
BC053855.1
-
9
2269
5588610
EU887632.1
-
10
2437
EU887633.1
-
10
2437
EU887634.1
-
10
2437
EU887635.1
-
10
2253
EU887636.1
-
10
2253
EU887637.1
-
10
2253
EU887638.1
-
10
2306
EU887639.1
-
10
2306
EU887640.1
-
10
2306
EU887641.1
-
9
2132
EU887642.1
-
9
2132
EU887643.1
-
9
2132
EU887644.1
-
9
2256
EU887645.1
-
9
2256
EU887646.1
-
9
2256
EU887647.1
-
8
2378
EU887648.1
-
8
2378
EU887649.1
-
8
2378
EU887650.1
-
6
1129
EU887651.1
-
6
1129
EU887652.1
-
6
1129
EU887653.1
-
4
603
EU887654.1
-
4
604
EU887655.1
-
4
603
L41066.1
-
9
2273
Back To Top
More Information
Panther Classification:
Molecular Function -
Rel homology transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
patterning of blood vessels
transcription from RNA polymerase II promoter
inflammatory response
heart development
intrinsic apoptotic signaling pathway in response to DNA damage
negative regulation of Wnt signaling pathway
negative regulation of protein binding
positive regulation of tumor necrosis factor production
calcineurin-NFAT signaling cascade
cellular response to UV
negative regulation of chromatin binding
cellular respiration
positive regulation of transcription from RNA polymerase II promoter
regulation of synaptic plasticity
negative regulation of dendrite morphogenesis
smooth muscle cell differentiation
muscle cell development
cellular response to lithium ion
negative regulation of pri-miRNA transcription from RNA polymerase II promoter
negative regulation of synapse maturation
positive regulation of apoptotic signaling pathway
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding
transcription coactivator activity
protein binding
transcription factor binding
peroxisome proliferator activated receptor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-4sktr:80/100.66.76.202:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0