Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NFE2L2 SIRNA Assays ›
          Gene Symbol:
          NFE2L2
          Gene Name:
          nuclear factor, erythroid 2 like 2
          Gene Aliases:
          HEBP1, NRF2
          Chromosome Location:
          Chr.2: 177230303 - 177265131 on Build GRCh38
          UniGene ID:
          Hs.744006
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS107130
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: NFE2L2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001145412.3 NP_001138884.1 5 1482
          NM_001145413.3 NP_001138885.1 5 1461
          NM_001313900.1 NP_001300829.1 5 1356
          NM_001313901.1 NP_001300830.1 5 1448
          NM_001313902.1 NP_001300831.1 4 1263
          NM_001313903.1 NP_001300832.1 5 1134
          NM_001313904.1 NP_001300833.1 5 1411
          NM_006164.4 NP_006155.2 5 1353
          GenBank mRNA AB208786.1 - 5 864
          AK297838.1 - 5 857
          AK304555.1 - 5 902
          AK311395.1 - 5 884
          AK314816.1 - 5 1354
          AK316148.1 - 5 805
          BC011558.1 - 5 911 4548874
          BX649047.1 - 5 892
          BX649071.1 - 4 693
          HM446346.1 - 5 805
          S74017.1 - 5 789

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          basic leucine zipper transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription from RNA polymerase II promoter
          inflammatory response
          proteasomal ubiquitin-independent protein catabolic process
          positive regulation of gene expression
          protein ubiquitination
          positive regulation of blood coagulation
          endoplasmic reticulum unfolded protein response
          cellular response to oxidative stress
          cellular response to drug
          positive regulation of transcription from RNA polymerase II promoter in response to stress
          positive regulation of transcription from RNA polymerase II promoter in response to oxidative stress
          PERK-mediated unfolded protein response
          cellular response to glucose starvation
          proteasome-mediated ubiquitin-dependent protein catabolic process
          cell redox homeostasis
          positive regulation of transcription from RNA polymerase II promoter
          regulation of embryonic development
          positive regulation of glucose import
          cellular response to hydrogen peroxide
          cellular response to tumor necrosis factor
          cellular response to fluid shear stress
          cellular response to laminar fluid shear stress
          negative regulation of hematopoietic stem cell differentiation
          negative regulation of oxidative stress-induced intrinsic apoptotic signaling pathway
          positive regulation of ER-associated ubiquitin-dependent protein catabolic process
          negative regulation of hydrogen peroxide-induced cell death
          positive regulation of glutathione biosynthetic process
          regulation of removal of superoxide radicals
          negative regulation of endothelial cell apoptotic process
          positive regulation of reactive oxygen species metabolic process
          transcription regulatory region sequence-specific DNA binding
          RNA polymerase II distal enhancer sequence-specific DNA binding
          RNA polymerase II activating transcription factor binding
          transcriptional activator activity, RNA polymerase II distal enhancer sequence-specific binding
          transcription cofactor binding
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          protein binding
          protein domain specific binding
          transcription regulatory region DNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0