Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PCDH7 SIRNA Assays ›
Gene Symbol:
PCDH7
Gene Name:
protocadherin 7
Gene Aliases:
BH-Pcdh, BHPCDH, PPP1R120
Chromosome Location:
Chr.4: 30720378 - 31146801 on Build GRCh38
UniGene ID:
Hs.479439
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS107644
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PCDH7
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001173523.1
NP_001166994.1
1
2254
NM_002589.2
NP_002580.2
1
2254
NM_032456.2
NP_115832.1
1
2261
NM_032457.3
NP_115833.2
1
2254
XM_005248163.3
XP_005248220.1
1
2289
XM_005248164.3
XP_005248221.1
1
2289
XM_005248166.3
XP_005248223.1
1
2288
XM_011513842.2
XP_011512144.1
1
2290
XM_011513843.2
XP_011512145.1
1
2289
XM_011513844.2
XP_011512146.1
1
2290
XM_017008259.1
XP_016863748.1
1
2290
XM_017008260.1
XP_016863749.1
1
2290
XM_017008261.1
XP_016863750.1
1
2290
XM_017008262.1
XP_016863751.1
1
2290
XM_017008263.1
XP_016863752.1
1
2291
XM_017008264.1
XP_016863753.1
1
2291
XM_017008265.1
XP_016863754.1
1
2291
XM_017008266.1
XP_016863755.1
1
2291
XM_017008267.1
XP_016863756.1
1
2291
XM_017008268.1
XP_016863757.1
1
2291
XM_017008269.1
XP_016863758.1
1
2291
XM_017008270.1
XP_016863759.1
1
2290
XM_017008271.1
XP_016863760.1
1
2290
XM_017008272.1
XP_016863761.1
1
2289
GenBank mRNA
AB006755.1
-
1
2256
AB006756.1
-
1
1813
AB006757.1
-
2
1960
Back To Top
More Information
Panther Classification:
Molecular Function -
cadherin
Gene Ontology Categories:
Function(s)
Process(es)
platelet degranulation
homophilic cell adhesion via plasma membrane adhesion molecules
calcium ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-wrkvw:80/100.66.78.186:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0