Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PDPK1 SIRNA Assays ›
          Gene Symbol:
          PDPK1
          Gene Name:
          3-phosphoinositide dependent protein kinase 1
          Gene Aliases:
          PDK1, PDPK2, PDPK2P, PRO0461
          Chromosome Location:
          Chr.16: 2537964 - 2603190 on Build GRCh38
          UniGene ID:
          Hs.459691
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS107791
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PDPK1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001261816.1 NP_001248745.1 11 1379
          NM_002613.4 NP_002604.1 11 1379
          NM_031268.5 NP_112558.2 8 998
          XM_011522521.2 XP_011520823.1 11 5067
          XM_011522523.2 XP_011520825.1 8 4691
          XM_017023271.1 XP_016878760.1 11 5072
          XM_017023272.1 XP_016878761.1 11 1628
          XM_017023273.1 XP_016878762.1 11 1337
          XM_017023274.1 XP_016878763.1 12 1475
          XM_017023275.1 XP_016878764.1 8 4697
          XM_017023276.1 XP_016878765.1 8 941
          XM_017023277.1 XP_016878766.1 9 1039
          GenBank mRNA AB209835.1 - 11 1346
          AF017995.1 - 11 1310
          AK131060.1 - 11 1313
          AK222581.1 - 11 1322
          BC006339.2 - 8 959 4079551
          BC012103.1 - 11 1339 4561371
          BC033494.1 - 8 911 5176294
          CR536517.1 - 11 1230
          Y15056.1 - 11 1309

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          stimulatory C-type lectin receptor signaling pathway
          type B pancreatic cell development
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          protein phosphorylation
          negative regulation of protein kinase activity
          hyperosmotic response
          epidermal growth factor receptor signaling pathway
          positive regulation of phospholipase activity
          regulation of endothelial cell migration
          negative regulation of cardiac muscle cell apoptotic process
          cell migration
          peptidyl-serine phosphorylation
          peptidyl-threonine phosphorylation
          calcium-mediated signaling
          actin cytoskeleton organization
          platelet activation
          negative regulation of transforming growth factor beta receptor signaling pathway
          T cell costimulation
          activation of protein kinase B activity
          cellular response to insulin stimulus
          negative regulation of toll-like receptor signaling pathway
          intracellular signal transduction
          Fc-epsilon receptor signaling pathway
          regulation of I-kappaB kinase/NF-kappaB signaling
          regulation of mast cell degranulation
          protein autophosphorylation
          phosphatidylinositol-mediated signaling
          focal adhesion assembly
          T cell receptor signaling pathway
          positive regulation of release of sequestered calcium ion into cytosol
          cellular response to epidermal growth factor stimulus
          positive regulation of establishment of protein localization to plasma membrane
          extrinsic apoptotic signaling pathway
          protein serine/threonine kinase activity
          3-phosphoinositide-dependent protein kinase activity
          insulin receptor binding
          protein binding
          ATP binding
          phospholipase activator activity
          kinase activity
          protein kinase binding
          phospholipase binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0