Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PFKM SIRNA Assays ›
Gene Symbol:
PFKM
Gene Name:
phosphofructokinase, muscle
Gene Aliases:
ATP-PFK, GSD7, PFK-1, PFK1, PFKA, PFKX, PPP1R122
Chromosome Location:
Chr.12: 48105278 - 48146404 on Build GRCh38
UniGene ID:
Hs.75160
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS107874
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Contact Us
Your Price
Online offer:
3062.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PFKM
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000289.5
NP_000280.1
17
1817
NM_001166686.1
NP_001160158.1
19
2119
NM_001166687.1
NP_001160159.1
17
1671
NM_001166688.1
NP_001160160.1
16
1760
XM_005268974.1
XP_005269031.1
20
2016
XM_005268975.1
XP_005269032.1
20
2155
XM_005268976.3
XP_005269033.1
20
2198
XM_005268977.1
XP_005269034.1
19
2002
XM_005268978.2
XP_005269035.1
19
1915
XM_005268979.1
XP_005269036.1
19
2002
XM_011538487.1
XP_011536789.1
18
1857
XM_011538488.2
XP_011536790.1
17
1782
XM_017019467.1
XP_016874956.1
20
2085
XM_017019468.1
XP_016874957.1
19
1937
XM_017019469.1
XP_016874958.1
18
1909
GenBank mRNA
AK126020.1
-
21
2472
AK126229.1
-
20
2069
AK303681.1
-
12
1164
AK308185.1
-
12
1140
AK312284.1
-
17
1699
BC000534.2
-
16
1647
2964710
BC007798.2
-
15
1534
BC012799.2
-
16
1647
2964710
BC013298.2
-
16
1647
2964710
BC021203.2
-
17
1669
4100117
BX537703.1
-
1
821
CR541698.1
-
7
687
J05533.1
-
8
728
KU178251.1
-
16
1632
KU178252.1
-
15
1539
KU178253.1
-
16
1632
KU178254.1
-
5
477
M26066.1
-
17
1708
U24183.1
-
16
1960
Y00698.1
-
16
1651
Back To Top
More Information
Panther Classification:
Molecular Function -
carbohydrate kinase
kinase
transferase
metabolite interconversion enzyme
Gene Ontology Categories:
Function(s)
Process(es)
glycogen catabolic process
fructose 6-phosphate metabolic process
glycolytic process
positive regulation of insulin secretion
glucose homeostasis
muscle cell cellular homeostasis
carbohydrate phosphorylation
protein oligomerization
canonical glycolysis
glycolysis from storage polysaccharide through glucose-1-phosphate
6-phosphofructokinase activity
protein binding
ATP binding
protein C-terminus binding
kinase binding
identical protein binding
protein homodimerization activity
metal ion binding
fructose binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-5thf6:80/100.66.79.225:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0