Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PKHD1 SIRNA Assays ›
          Gene Symbol:
          PKHD1
          Gene Name:
          polycystic kidney and hepatic disease 1 (autosomal recessive)
          Gene Aliases:
          ARPKD, FCYT, TIGM1
          Chromosome Location:
          Chr.6: 51613291 - 52087625 on Build GRCh38
          UniGene ID:
          Hs.662050
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS108049
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PKHD1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_138694.3 NP_619639.3 35 5991
          NM_170724.2 NP_733842.2 35 5991
          XM_011514680.2 XP_011512982.1 35 6335
          XM_011514682.2 XP_011512984.1 35 5957
          XM_011514683.2 XP_011512985.1 36 5315
          XM_011514684.2 XP_011512986.1 27 5101
          XM_011514685.1 XP_011512987.1 35 5991
          XM_011514686.2 XP_011512988.1 35 5991
          XM_011514687.1 XP_011512989.1 35 5991
          XM_011514688.2 XP_011512990.1 35 5957
          XM_011514690.2 XP_011512992.1 1 34
          XM_017010944.1 XP_016866433.1 35 5817
          XM_017010945.1 XP_016866434.1 34 5882
          XM_017010946.1 XP_016866435.1 35 5957
          XM_017010947.1 XP_016866436.1 33 5693
          XM_017010948.1 XP_016866437.1 26 5775
          XM_017010949.1 XP_016866438.1 17 4954
          XM_017010950.1 XP_016866439.1 35 5991
          XM_017010951.1 XP_016866440.1 35 5957
          XM_017010952.1 XP_016866441.1 35 5957
          GenBank mRNA AF480064.1 - 35 5937
          AY074797.1 - 35 5991
          AY092083.1 - 35 5977

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          kidney development
          cellular calcium ion homeostasis
          positive regulation of cell proliferation
          regulation of centrosome duplication
          single organismal cell-cell adhesion
          regulation of TOR signaling
          negative regulation of NF-kappaB transcription factor activity
          cilium assembly
          homeostatic process
          negative regulation of apoptotic process
          negative regulation of cellular component movement
          negative regulation of protein kinase B signaling
          regulation of ERK1 and ERK2 cascade
          receptor activity
          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0