Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PKHD1 SIRNA Assays ›
Gene Symbol:
PKHD1
Gene Name:
polycystic kidney and hepatic disease 1 (autosomal recessive)
Gene Aliases:
ARPKD, FCYT, TIGM1
Chromosome Location:
Chr.6: 51613291 - 52087625 on Build GRCh38
UniGene ID:
Hs.662050
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS108051
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PKHD1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_138694.3
NP_619639.3
45, 46
7481
NM_170724.2
NP_733842.2
45, 46
7481
XM_011514680.2
XP_011512982.1
45, 46
7825
XM_011514682.2
XP_011512984.1
45, 46
7447
XM_011514683.2
XP_011512985.1
46, 47
6805
XM_011514684.2
XP_011512986.1
37, 38
6591
XM_011514685.1
XP_011512987.1
45, 46
7481
XM_011514686.2
XP_011512988.1
45, 46
7481
XM_011514687.1
XP_011512989.1
45, 46
7481
XM_011514688.2
XP_011512990.1
45, 46
7447
XM_011514690.2
XP_011512992.1
10, 11
1367
XM_011514691.2
XP_011512993.1
10, 11
7392
XM_017010944.1
XP_016866433.1
45, 46
7307
XM_017010945.1
XP_016866434.1
44, 45
7372
XM_017010946.1
XP_016866435.1
45, 46
7447
XM_017010947.1
XP_016866436.1
43, 44
7183
XM_017010948.1
XP_016866437.1
36, 37
7265
XM_017010949.1
XP_016866438.1
27, 28
6444
XM_017010950.1
XP_016866439.1
45, 46
7481
XM_017010951.1
XP_016866440.1
45, 46
7447
XM_017010952.1
XP_016866441.1
45, 46
7447
GenBank mRNA
AF480064.1
-
45, 46
7427
AK128031.1
-
9, 10
1404
AY074797.1
-
45, 46
7481
AY092083.1
-
45, 46
7467
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
kidney development
cellular calcium ion homeostasis
positive regulation of cell proliferation
regulation of centrosome duplication
single organismal cell-cell adhesion
regulation of TOR signaling
negative regulation of NF-kappaB transcription factor activity
cilium assembly
homeostatic process
negative regulation of apoptotic process
negative regulation of cellular component movement
negative regulation of protein kinase B signaling
regulation of ERK1 and ERK2 cascade
receptor activity
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0