Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PPARD SIRNA Assays ›
          Gene Symbol:
          PPARD
          Gene Name:
          peroxisome proliferator activated receptor delta
          Gene Aliases:
          FAAR, NR1C2, NUC1, NUCI, NUCII, PPARB
          Chromosome Location:
          Chr.6: 35342558 - 35428191 on Build GRCh38
          UniGene ID:
          Hs.696032
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS108294
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PPARD
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001171818.1 NP_001165289.1 7 955
          NM_001171819.1 NP_001165290.1 5 628
          NM_001171820.1 NP_001165291.1 4 565
          NM_006238.4 NP_006229.1 6 859
          NM_177435.2 NP_803184.1 6 859
          XM_005249193.1 XP_005249250.1 6 874
          XM_006715120.1 XP_006715183.1 8 1012
          XM_006715123.1 XP_006715186.1 5 762
          XM_011514707.1 XP_011513009.1 8 1609
          XM_011514709.1 XP_011513011.1 8 1583
          XM_011514710.1 XP_011513012.1 7 936
          XM_017010971.1 XP_016866460.1 9 1705
          XM_017010972.1 XP_016866461.1 8 1636
          XM_017010973.1 XP_016866462.1 8 1071
          XM_017010974.1 XP_016866463.1 7 996
          XM_017010975.1 XP_016866464.1 5 747
          GenBank mRNA AB307691.1 - 4 550
          AK122614.1 - 6 852
          AK291661.1 - 6 818
          AK296425.1 - 5 587
          AK304878.1 - 4 528
          AY919140.1 - 4 550
          BC002715.2 - 6 808 3630487
          BC007578.1 - 6 808 3139849
          CN332566.1 - 3 422
          HQ692865.1 - 4 550
          L07592.1 - 7 887

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_013141, XM_006256168, XM_008772797, XM_006256166, XM_006256167, and others.

          Panther Classification:

          Molecular Function -

          C4 zinc finger nuclear receptor DNA-binding transcription factor zinc finger transcription factor gene-specific transcriptional regulator

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          glucose metabolic process
          proteoglycan metabolic process
          generation of precursor metabolites and energy
          regulation of transcription from RNA polymerase II promoter
          transcription initiation from RNA polymerase II promoter
          lipid metabolic process
          fatty acid beta-oxidation
          vitamin A metabolic process
          apoptotic process
          heart development
          embryo implantation
          cholesterol metabolic process
          cell proliferation
          axon ensheathment
          phospholipid biosynthetic process
          fatty acid catabolic process
          mRNA transcription
          response to glucose
          positive regulation of phosphatidylinositol 3-kinase signaling
          response to activity
          regulation of skeletal muscle satellite cell proliferation
          negative regulation of smooth muscle cell migration
          glucose transport
          fatty acid transport
          cell differentiation
          negative regulation of cell growth
          intracellular receptor signaling pathway
          cell-substrate adhesion
          positive regulation of insulin secretion
          negative regulation of collagen biosynthetic process
          response to vitamin A
          wound healing
          negative regulation of apoptotic process
          steroid hormone mediated signaling pathway
          positive regulation of skeletal muscle tissue regeneration
          keratinocyte proliferation
          positive regulation of fat cell differentiation
          negative regulation of myoblast differentiation
          positive regulation of epidermis development
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          positive regulation of vasodilation
          decidualization
          negative regulation of smooth muscle cell proliferation
          negative regulation of epithelial cell proliferation
          negative regulation of inflammatory response
          keratinocyte migration
          adipose tissue development
          cellular response to lipopolysaccharide
          cellular response to hypoxia
          apoptotic signaling pathway
          negative regulation of pri-miRNA transcription from RNA polymerase II promoter
          positive regulation of myoblast proliferation
          transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          steroid hormone receptor activity
          transcription coactivator activity
          RNA polymerase II transcription factor activity, ligand-activated sequence-specific DNA binding
          protein binding
          transcription factor binding
          drug binding
          zinc ion binding
          lipid binding
          sequence-specific DNA binding
          protein heterodimerization activity
          NF-kappaB binding
          linoleic acid binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-4x8c5:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0