Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PPP5C SIRNA Assays ›
          Gene Symbol:
          PPP5C PPP5D1
          Gene Name:
          protein phosphatase 5 catalytic subunit
          PPP5 tetratricopeptide repeat domain containing 1
          Gene Aliases:
          PP5, PPP5, PPT
          -
          Chromosome Location:
          Chr.19: 46346994 - 46390975 on Build GRCh38
          Chromosome Location:
          Chr.19: 46480788 - 46601200 on Build GRCh38
          UniGene ID:
          Hs.654604
          Hs.671661
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID HSS108417
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PPP5C
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001204284.1 NP_001191213.1 3 525
          NM_006247.3 NP_006238.1 3 525
          XM_017026935.1 XP_016882424.1 3 520
          GenBank mRNA AK223222.1 - 3 432
          AK293402.1 - 4 383
          AK297095.1 - 4 371
          AK312683.1 - 3 432
          BC000750.2 - 3 418
          BC001831.2 - 3 418
          BC001970.1 - 3 482 3459309
          BT007275.1 - 3 422
          DA545760.1 - 3 526
          U25174.1 - 3 400
          X89416.1 - 3 405
          Gene Symbol: PPP5D1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001205281.1 NP_001192210.1 2 377
          XM_005258397.3 XP_005258454.1 2 377
          XM_006722960.3 XP_006723023.1 2 377
          XM_017026125.1 XP_016881614.1 2 377
          GenBank mRNA AK094504.1 - 2 377
          BM718195.1 - 2 109
          DA130492.1 - 2 377

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          protein phosphatase

          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          negative regulation of protein phosphorylation
          DNA repair
          transcription, DNA-templated
          protein dephosphorylation
          mitotic nuclear division
          response to lead ion
          histone dephosphorylation
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          response to morphine
          protein heterooligomerization
          cellular response to hydrogen peroxide
          cellular response to cadmium ion
          negative regulation of neuron death
          positive regulation of glucocorticoid receptor signaling pathway
          G-protein alpha-subunit binding
          RNA binding
          phosphoprotein phosphatase activity
          protein serine/threonine phosphatase activity
          signal transducer activity
          protein binding
          ATP binding
          microtubule binding
          lipid binding
          heat shock protein binding
          identical protein binding
          ADP binding
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-x88k6:80/100.66.75.98:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0