Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PRKCE SIRNA Assays ›
          Gene Symbol:
          PRKCE
          Gene Name:
          protein kinase C epsilon
          Gene Aliases:
          PKCE, nPKC-epsilon
          Chromosome Location:
          Chr.2: 45651315 - 46187990 on Build GRCh38
          UniGene ID:
          Hs.580351
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS108501
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PRKCE
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_005400.2 NP_005391.1 6 1004
          XM_005264428.1 XP_005264485.1 7 1033
          XM_005264431.3 XP_005264488.1 6 1593
          XM_011532971.2 XP_011531273.1 8 1657
          XM_011532975.2 XP_011531277.1 10 1145
          XM_011532978.1 XP_011531280.1 7 637
          XM_011532980.2 XP_011531282.1 7 674
          XM_011532981.2 XP_011531283.1 5 1797
          XM_011532982.1 XP_011531284.1 4 809
          XM_017004486.1 XP_016859975.1 8 1699
          XM_017004487.1 XP_016859976.1 8 746
          XM_017004488.1 XP_016859977.1 7 1481
          XM_017004489.1 XP_016859978.1 11 1086
          XM_017004490.1 XP_016859979.1 7 1586
          XM_017004491.1 XP_016859980.1 6 637
          XM_017004492.1 XP_016859981.1 6 1593
          GenBank mRNA AK313842.1 - 6 903
          AL832345.1 - 6 1134
          BC109033.2 - 6 942 40028213
          BC109034.2 - 6 942 40028215
          X65293.1 - 6 822

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase protein modifying enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          macrophage activation involved in immune response
          protein phosphorylation
          apoptotic process
          cell cycle
          cell adhesion
          signal transduction
          activation of phospholipase C activity
          positive regulation of epithelial cell migration
          positive regulation of fibroblast migration
          positive regulation of cell-substrate adhesion
          peptidyl-serine phosphorylation
          platelet activation
          positive regulation of actin filament polymerization
          negative regulation of protein ubiquitination
          lipopolysaccharide-mediated signaling pathway
          positive regulation of insulin secretion
          positive regulation of synaptic transmission, GABAergic
          positive regulation of cytokinesis
          intracellular signal transduction
          locomotory exploration behavior
          TRAM-dependent toll-like receptor 4 signaling pathway
          Fc-gamma receptor signaling pathway involved in phagocytosis
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          response to morphine
          positive regulation of MAPK cascade
          regulation of peptidyl-tyrosine phosphorylation
          positive regulation of lipid catabolic process
          release of sequestered calcium ion into cytosol
          regulation of release of sequestered calcium ion into cytosol
          cell division
          regulation of insulin secretion involved in cellular response to glucose stimulus
          positive regulation of mucus secretion
          cellular response to ethanol
          cellular response to prostaglandin E stimulus
          cellular response to hypoxia
          positive regulation of wound healing
          positive regulation of receptor activity
          negative regulation of sodium ion transmembrane transporter activity
          positive regulation of cellular glucuronidation
          actin monomer binding
          protein serine/threonine kinase activity
          protein kinase C activity
          calcium-independent protein kinase C activity
          signal transducer activity
          protein binding
          ATP binding
          enzyme activator activity
          enzyme binding
          receptor activator activity
          ethanol binding
          metal ion binding
          14-3-3 protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0