Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PTGER4 SIRNA Assays ›
          Gene Symbol:
          PTGER4
          Gene Name:
          prostaglandin E receptor 4
          Gene Aliases:
          EP4, EP4R
          Chromosome Location:
          Chr.5: 40679930 - 40740932 on Build GRCh38
          UniGene ID:
          Hs.199248
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS108770
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PTGER4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000958.2 NP_000949.1 3 1578
          XM_017009656.1 XP_016865145.1 3 1121
          XM_017009657.1 XP_016865146.1 3 1087
          GenBank mRNA AY429109.1 - 2 986
          BC101534.1 - 3 1045 8069040
          BC113523.1 - 3 1045 8322575
          D28472.1 - 3 1070
          L25124.1 - 3 1571
          L28175.1 - 3 1374

          Back To Top

          More Information


          Set Membership:

          G-protein coupled receptor

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of peptide secretion
          immune response
          adenylate cyclase-modulating G-protein coupled receptor signaling pathway
          adenylate cyclase-activating G-protein coupled receptor signaling pathway
          positive regulation of cytosolic calcium ion concentration
          JNK cascade
          female pregnancy
          positive regulation of cell proliferation
          response to mechanical stimulus
          positive regulation of gene expression
          negative regulation of hydrogen peroxide metabolic process
          regulation of circadian sleep/wake cycle, wakefulness
          positive regulation of smooth muscle cell migration
          positive regulation of cAMP biosynthetic process
          response to lipopolysaccharide
          response to progesterone
          positive regulation of interleukin-8 production
          negative regulation of integrin activation
          positive regulation of urine volume
          positive regulation of renal sodium excretion
          T-helper cell differentiation
          negative regulation of circadian sleep/wake cycle, REM sleep
          chemokinesis
          response to drug
          positive regulation of tyrosine phosphorylation of Stat3 protein
          response to amino acid
          positive regulation of osteoblast differentiation
          positive regulation of bone resorption
          positive regulation of cell adhesion
          positive regulation of circadian sleep/wake cycle, non-REM sleep
          negative regulation of cytokine secretion
          negative regulation of interleukin-1 alpha secretion
          positive regulation of cytokine secretion
          negative regulation of inflammatory response
          positive regulation of inflammatory response
          regulation of stress fiber assembly
          negative regulation of nitric-oxide synthase biosynthetic process
          maternal process involved in parturition
          bone development
          positive regulation of mucus secretion
          ERK1 and ERK2 cascade
          cellular response to mechanical stimulus
          cellular response to glucose stimulus
          cellular response to interleukin-1
          cellular response to prostaglandin E stimulus
          positive regulation of wound healing
          ductus arteriosus closure
          positive regulation of hyaluronan biosynthetic process
          response to salt
          negative regulation of ductus arteriosus closure
          negative regulation of small intestine smooth muscle contraction
          positive regulation of calcitonin secretion
          positive regulation of chemokinesis
          positive regulation of matrix metallopeptidase secretion
          negative regulation of tumor necrosis factor secretion
          negative regulation of endothelin secretion
          positive regulation of substance P secretion, neurotransmission
          response to water-immersion restraint stress
          positive regulation of antral ovarian follicle growth
          positive regulation of neutrophil extravasation
          negative regulation of eosinophil extravasation
          positive regulation of interleukin-10 secretion
          prostaglandin E receptor activity
          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0