Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PXN SIRNA Assays ›
Gene Symbol:
PXN
Gene Name:
paxillin
Gene Aliases:
-
Chromosome Location:
Chr.12: 120210439 - 120265771 on Build GRCh38
UniGene ID:
Hs.446336
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS108925
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Contact Us
Your Price
Online offer:
3062.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PXN
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001080855.2
NP_001074324.1
12
1885
NM_001243756.1
NP_001230685.1
11
1927
NM_002859.3
NP_002850.2
11
1783
NM_025157.4
NP_079433.3
11
1907
XM_005253917.3
XP_005253974.1
7
1282
XM_006719532.1
XP_006719595.1
15
3468
XM_006719534.1
XP_006719597.1
15
3623
XM_006719535.3
XP_006719598.1
16
3551
XM_006719536.2
XP_006719599.1
15
3440
XM_011538622.1
XP_011536924.1
16
3550
XM_011538623.1
XP_011536925.1
16
3737
XM_017019726.1
XP_016875215.1
15
4036
XM_017019727.1
XP_016875216.1
15
3836
XM_017019728.1
XP_016875217.1
15
3827
XM_017019729.1
XP_016875218.1
15
3892
XM_017019731.1
XP_016875220.1
16
3921
XM_017019733.1
XP_016875222.1
13
3451
XM_017019734.1
XP_016875223.1
15
3348
XM_017019735.1
XP_016875224.1
12
3205
XM_017019736.1
XP_016875225.1
13
2875
XM_017019737.1
XP_016875226.1
13
2731
XM_017019738.1
XP_016875227.1
12
2566
XM_017019739.1
XP_016875228.1
11
2464
XM_017019740.1
XP_016875229.1
12
2422
XM_017019741.1
XP_016875230.1
12
2213
XM_017019742.1
XP_016875231.1
11
2320
XM_017019743.1
XP_016875232.1
12
2021
GenBank mRNA
AB209034.1
-
14
3561
AK122964.1
-
12
2213
AK128712.1
-
9
2233
AK299486.1
-
9
1214
AK314204.1
-
11
1742
BC136787.1
-
11
1717
9020777
BC136794.1
-
11
1717
9020784
BC144410.1
-
12
1819
9052931
BM742662.1
-
1
116
BX648777.1
-
11
1901
D86862.1
-
11
1743
D86863.1
-
10
1785
U14588.1
-
11
1715
Back To Top
More Information
Panther Classification:
Molecular Function -
actin or actin-binding cytoskeletal protein
Gene Ontology Categories:
Function(s)
Process(es)
muscle contraction
cell adhesion
cell-matrix adhesion
signal transduction
signal complex assembly
epidermal growth factor receptor signaling pathway
transforming growth factor beta receptor signaling pathway
cellular response to reactive oxygen species
vascular endothelial growth factor receptor signaling pathway
growth hormone receptor signaling pathway
integrin binding
protein binding
beta-catenin binding
zinc ion binding
vinculin binding
protein kinase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-fz5f2:80/100.66.72.225:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0